Skip to Content
Merck
All Photos(1)

Documents

703753

Sigma-Aldrich

Di-tert-butyl dicarbonate solution

2 M in THF

Synonym(s):

Di-tert-butyl pyrocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O[CO2C(CH3)3]2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2 M in THF

refractive index

n20/D 1.408

density

0.934 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

InChI

1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3

InChI key

DYHSDKLCOJIUFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans Peter Reisenauer et al.
Angewandte Chemie (International ed. in English), 53(44), 11766-11771 (2014-09-10)
Carbonic acid (H2CO3), an essential molecule of life (e.g., as bicarbonate buffer), has been well characterized in solution and in the solid state, but for a long time, it has eluded its spectral characterization in the gas phase owing to
E E Büllesbach et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 361(5), 723-734 (1980-05-01)
Nepsilon 29, Nepsilon 59-Bis(methylsulfonylethoxycarbonyl)proinsulin was prepared from native beef proinsulin for its possible use in the semisynthesis of preproinsulin. The modification was made possible by first protecting the alpha-amino group of proinsulin either with the tert-butoxycarbonyl group, after reaction with
Asit K Chakraborti et al.
Organic & biomolecular chemistry, 4(14), 2769-2771 (2006-07-11)
Perchloric acid adsorbed on silica-gel (HClO4-SiO2) was found to be a new, highly efficient, inexpensive and reusable catalyst for chemoselective N-tert-butoxycarbonylation of amines at room temperature and under solvent-free conditions.
Jason C Green et al.
Organic letters, 13(20), 5500-5503 (2011-09-21)
Total syntheses of two structures purported as (+)-heliananes were completed in six pots. Spectral comparisons, between the synthetic and natural compounds, revealed a misassignment of the eight-membered ring in the heliananes. The key step in the syntheses of the proposed
Brandon S Fowler et al.
Journal of the American Chemical Society, 132(9), 2870-2871 (2010-02-18)
We report a fundamentally unique approach to the catalytic kinetic resolution of amine derivatives based on formamide and thioformamide substrates. Readily accessible histidine-containing peptides mediate the kinetic resolutions with as little as 5 mol % catalyst. Selectivity factors (k(rel)) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service