Skip to Content
Merck
Di-tert-butyl dicarbonate solution 2 M in methylene chloride

Di-tert-butyl dicarbonate solution

Di-tert-butyl dicarbonate solution 2 M in methylene chloride
Synonym(s):
Di-tert-butyl pyrocarbonate
Linear Formula:
O[CO2C(CH3)3]2
CAS Number:
Molecular Weight:
218.25

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group