Skip to Content
Merck
All Photos(1)

Documents

148504

Sigma-Aldrich

Triphenylsilane

97%

Synonym(s):

1,1′,1′′-Silylidynetrisbenzene, NSC 12565, Triphenylhydrosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SiH
CAS Number:
Molecular Weight:
260.41
Beilstein:
978182
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: reductant

bp

152 °C/2 mmHg (lit.)

mp

43-45 °C (lit.)

SMILES string

c1ccc(cc1)[SiH](c2ccccc2)c3ccccc3

InChI

1S/C18H16Si/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H

InChI key

AKQNYQDSIDKVJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent:
  • For catalytic hydrogen deuterium exchange reactions of silanes
  • To be oxidized by carbon nanotube-gold nanohybrids
  • For hydrolysis by ruthenium complexes
  • For hydrosilylation to produce enolsilanes
  • For synthesis of bromosilanes
  • For ozone oxidation of silyl-alkenes for synthesis of α-O-silylated acyloin derivatives
  • Used to synthesize hydrido silyl and bis(silyl) bis(imidazolinylidene) nickel complexes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sylwia Członka et al.
Materials (Basel, Switzerland), 13(7) (2020-04-16)
In this work, rigid polyurethane (PUR) foams were prepared by incorporating 2 wt% of eucalyptus fibers. The eucalyptus fibers were surface-modified by maleic anhydride, alkali, and silane (triphenylsilanol) treatment. The impact of the modified eucalyptus fibers on the mechanical, thermal
Courtney M Olson et al.
The Journal of chemical physics, 147(12), 124302-124302 (2017-10-02)
Fourier transform infrared and two-dimensional IR (2D-IR) spectroscopies were applied to two different silanes in three different solvents. The selected solutes exhibit different degrees of vibrational solvatochromism for the Si-H vibration. Density functional theory calculations confirm that this difference in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service