Skip to Content
Merck
All Photos(1)

Documents

687529

Sigma-Aldrich

Poly(ethylene glycol) dimethacrylate

average MN 2,000, cross-linking reagent polymerization reactions, methacrylate, ~1000 ppm MeHQ as stabilizer

Synonym(s):

Polyethylene glycol, PEG dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer

form

powder

mol wt

average Mn 2000

contains

~1000 ppm MeHQ as stabilizer

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

bp

>200 °C/2 mmHg (lit.)

transition temp

Tm 49-55 °C

Mw/Mn

<1.2

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

OCCO.CC(=C)C(O)=O

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanhua Huang et al.
Carbohydrate polymers, 97(2), 496-501 (2013-08-06)
A novel biodegradable β-cyclodextrin-based gel (CAM) was prepared and applied to the removal of Cd(2+), Pb(2+) and Cu(2+) ions from aqueous solutions. CAM hydrogel has a typical three-dimensional network structure, and showed excellent capability for the removal of heavy metal
Hiroaki Onoe et al.
Nature materials, 12(6), 584-590 (2013-04-02)
Artificial reconstruction of fibre-shaped cellular constructs could greatly contribute to tissue assembly in vitro. Here we show that, by using a microfluidic device with double-coaxial laminar flow, metre-long core-shell hydrogel microfibres encapsulating ECM proteins and differentiated cells or somatic stem
Enrique Barragan Garza et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(3), 166-172 (2013-03-01)
To conduct a feasibility study of the safety and efficacy of a corneal contouring inlay as a treatment for emmetropic presbyopia. The Raindrop corneal inlay (ReVision Optics, Inc., Lake Forest, CA) was implanted on the corneal stromal bed beneath a
Kwanghun Chung et al.
Nature methods, 10(6), 508-513 (2013-06-01)
With potential relevance for brain-mapping work, hydrogel-based structures can now be built from within biological tissue to allow subsequent removal of lipids without mechanical disassembly of the tissue. This process creates a tissue-hydrogel hybrid that is physically stable, that preserves
Svenja Hinderer et al.
Scientific reports, 8(1), 110-110 (2018-01-10)
Decorin (DCN) is an important small leucine-rich proteoglycan present in the extracellular matrix (ECM) of many organs and tissues. Endothelial progenitor cells (EPCs) are able to interact with the surrounding ECM and bind to molecules such as DCN. Here, we

Articles

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Highlighting new synthetic modifications of PEG to improve the mechanical properties and degradation of resulting hydrogels in tissue engineering applications.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service