Skip to Content
Merck
All Photos(1)

Key Documents

475629

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average Mn 250, acrylate, 100 ppm MEHQ as inhibitor

Synonym(s):

Polyethylene glycol, PEG diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) diacrylate, average Mn 250

mol wt

average Mn 250

contains

100 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.463

density

1.11 g/mL at 25 °C

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

OCCO.OC(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) diacrylate (PEGDA) is a polyethylene glycol (PEG) based material that can be used for a variety of tissue engineering and drug delivery based applications. It is majorly used as a prepolymer solution that can be used in the formation of a cross-linked polymeric system.

Application

PEGDA can be used in the formation of a UV-cured membrane for potential usage in the separation of carbon dioxide (CO2) based gases. It may also be used in the development of novel injectable biodegradable polymers for a variety of biomedical applications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional thiols as additives in UV-cured PEG-diacrylate membranes for CO2 separation
Kwisnek L, et al.
Journal of Membrane Science , 369(1-2), 429-436 (2011)
Injectable biodegradable polymer composites based on poly (propylene fumarate) crosslinked with poly (ethylene glycol)-dimethacrylate
He S, et al.
Biomaterials, 21(23), 2389-2394 (2000)
Release of protein from highly cross-linked hydrogels of poly (ethylene glycol) diacrylate fabricated by UV polymerization
Mellott MB, et al.
Biomaterials, 22(9), 929-941 (2001)
Preparation and characterization of crosslinked poly (ethylene glycol) diacrylate hydrogels as fouling-resistant membrane coating materials
Ju H, et al.
Journal of Membrane Science , 330(1-2), 180-188 (2009)
Dibakar Mondal et al.
Journal of the mechanical behavior of biomedical materials, 104, 103653-103653 (2020-03-17)
In this study, single filaments of acrylated epoxidized soybean oil (AESO)/polyethylene glycol diacrylate (PEGDA)/nanohydroxyapatite (nHA)-based nanocomposites intended for bone defect repair have displayed significant improvement of their mechanical properties when extruded through smaller needle gauges before UV curing. These nanocomposite

Articles

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service