Skip to Content
Merck
All Photos(1)

Documents

62420

Sigma-Aldrich

Lithium aluminum hydride

≥97.0% (gas-volumetric)

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (gas-volumetric)

form

solid

reaction suitability

reagent type: reductant

mp

125 °C (dec.) (lit.)

SMILES string

[Li].[AlH3]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium aluminum hydride (LiAlH4) is a powerful reducing agent used in organic synthesis to reduce esters, carboxylic acids, acyl chlorides, aldehydes, epoxides, and ketones into the corresponding alcohols. It also converts amide, nitro, nitrile, imine, oxime, and azide compounds into amines.

Application

Reactant involved in:
  • Dehydrogenation studied with various catalysts
  • Regeneration of methoxysilanes via reduction
  • Transition metal electrophilic hydrogen activation via heterolytic cleavage
  • Formation of nickel hydrogenation catalysts
  • Hydrogen sorption
  • Electrodeposition of Al on W-Cu substrates

Packaging

packed in plastic bag, which is soluble at 45-60°C in hydrocarbons, THF and dioxane

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Q. Chen, et al.,
Surface and Coating Tech., 205, 4418-4424 (2011)
R. A. Varin and L. Zbroniec,
J. Alloy Compounds, 509, S736-S739 (2011)
Chu Liang et al.
Nature communications, 12(1), 119-119 (2021-01-07)
Environmentally benign synthesis of graphite at low temperatures is a great challenge in the absence of transition metal catalysts. Herein, we report a green and efficient approach of synthesizing graphite from carbon dioxide at ultralow temperatures in the absence of
Rafi-ud-din, et al.,
The Journal of Physical Chemistry C, 115, 13088-13099 (2011)
J. W. Runyon, et al.,
Australian Journal of Chemistry, 64, 1165-1172 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service