Skip to Content
Merck
All Photos(2)

Documents

212792

Sigma-Aldrich

Lithium aluminum hydride solution

1.0 M in diethyl ether

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in diethyl ether

density

0.716 g/mL at 25 °C

storage temp.

2-30°C

SMILES string

[Li+].[AlH4-]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for:
  • The preparation of thermoplastic polyester polyamides from oleic acid
  • Lithium-polymer batteries
  • Hydrodefluorination of gem-difluoromethylene derivatives
  • Asymmetric aldol reactions
  • Synthesis of Li-Al-N-H composites with hydrogen absorption / desorption properties

LAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcohols

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Yao et al.
Journal of electron microscopy, 60(6), 375-378 (2011-10-05)
A plastic bag method was developed to observe air-sensitive samples on microstructure and phase distribution without exposure to air during the holder transfer process into the transmission electron microscope (TEM). As an example, a type of lithium aluminum hydride (Li(3)AlH(6))
José M Concellón et al.
The Journal of organic chemistry, 71(17), 6420-6426 (2006-08-12)
The reaction of chiral (2R,1'S)- or (2S,1'S)-2-(1-aminoalkyl)epoxides, 1 or 2 with a variety of organolithium compounds to obtain the corresponding (alphaS,betaS)- or (alphaR,betaS)- beta-amino alcohols in enantiopure form is reported. In both cases, the opening of the oxirane ring at
I-Chi Lee et al.
Organic & biomolecular chemistry, 9(22), 7655-7658 (2011-09-17)
Specific deuterated reference compounds were prepared to probe the stereoselectivity of the reductive ring opening of carbohydrate-based benzylidene-type acetals. AlD(3) revealed a retentive stereoselectivity probably through the rare S(N)i (internal nucleophilic substitution) mechanism. An S(N)1-like mechanism occurs in the acid-promoted
Matthias D'hooghe et al.
Organic & biomolecular chemistry, 8(3), 607-615 (2010-01-22)
trans- and cis-1-Alkyl-4-aryl-3-chloroazetidin-2-ones, prepared through cyclocondensation of chloroketene and the appropriate imines in a diastereoselective way, were transformed into the corresponding non-activated trans- and cis-2-aryl-3-(hydroxymethyl)aziridines via reductive ring contraction using LiAlH(4) in Et(2)O. Furthermore, trans-2-aryl-3-(hydroxymethyl)aziridines were transformed into 2-amino-3-arylpropan-1-ols and
Pseudo-allosteric recognition of mandelic acid with an enantioselective coordination complex.
Jungseok Heo et al.
Angewandte Chemie (International ed. in English), 45(6), 941-944 (2005-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service