Skip to Content
Merck
All Photos(2)

Key Documents

554065

Sigma-Aldrich

Ethyl α-bromophenylacetate

97%

Synonym(s):

α-Bromobenzeneacetic acid ethyl ester, α-Bromophenylacetic acid ethyl ester, 2-Bromo-2-phenylacetic acid ethyl ester, Ethyl (±)-α-bromobenzeneacetate, Ethyl 2-bromo-2-phenylacetate, Ethyl 2-bromophenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHBrCOOCH2CH3
CAS Number:
Molecular Weight:
243.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D >1.5390 (lit.)

bp

89-92 °C/0.9 mmHg (lit.)

density

1.389 g/mL at 25 °C (lit.)

functional group

bromo
ester
phenyl

SMILES string

CCOC(=O)C(Br)c1ccccc1

InChI

1S/C10H11BrO2/c1-2-13-10(12)9(11)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3

InChI key

BKTKLDMYHTUESO-UHFFFAOYSA-N

Related Categories

Application

Ethyl α-bromophenylacetate (EBPA) may be used to synthesize α-ethoxycarbonyl-N,α-diphenylnitrone.
EBPA may be used as an initiator for the following:
  • polymerization of dimethyl(methacryloyloxymethyl) phosphonate
  • polymerization of methyl methacrylate (MMA)
  • polymerization of trimethylolpropane triacrylate(TMPTA)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence self-quenching of substituted N, a-diphenylnitrones in various solvents.
Khoee S and Memarian HR.
Journal of Photochemistry and Photobiology A: Chemistry, 177(2), 276-285 (2006)
Activators regenerated by electron transfer for atom-transfer radical polymerization of (meth)acrylates and related block copolymers.
Wojciech Jakubowski et al.
Angewandte Chemie (International ed. in English), 45(27), 4482-4486 (2006-06-14)
"A dinuclear gold (I) complex as a novel photoredox catalyst for light-induced atom transfer radical polymerization"
Nzulu F, et al.
Polym. Chem., 6(25), 4605-4611 (2015)
Wei Jiang et al.
Journal of colloid and interface science, 521, 62-68 (2018-03-20)
Atom transfer radical polymerization (ATRP) has been considered to be an efficient strategy for constructing functional macromolecules owing to its simple operation and versatile monomers, and thus it is of great significance to develop ideal catalysts with higher activity and
Control of a living radical polymerization of methacrylates by light.
Brett P Fors et al.
Angewandte Chemie (International ed. in English), 51(35), 8850-8853 (2012-07-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service