Skip to Content
Merck
All Photos(1)

Key Documents

369497

Sigma-Aldrich

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

99%

Synonym(s):

PMDETA, PMDTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NCH2CH2]2NCH3
CAS Number:
Molecular Weight:
173.30
Beilstein:
1741396
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.23 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

311 °F

expl. lim.

5.6 %

refractive index

n20/D 1.442 (lit.)

bp

198 °C (lit.)

mp

−20 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCN(C)CCN(C)C

InChI

1S/C9H23N3/c1-10(2)6-8-12(5)9-7-11(3)4/h6-9H2,1-5H3

InChI key

UKODFQOELJFMII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine (PMDETA) is an amine-based monomer compound that plays a crucial role in the field of polymers, particularly in the synthesis of TRPs- thermoresponsive polymers. PMDETA acts as a versatile ligand or catalyst in various polymerization reactions. It forms complexes with transition metals, which are utilized in the production of specialty polymers, block copolymers, and polymers with modified properties. These polymers find applications across a broad range of industries, including thermoplastics, elastomers, and coatings. When incorporated into the polymer structure, PMDETA helps impart thermoresponsive behavior to the final product, enabling reversible changes in solubility, conformation, or other physical properties in response to temperature variations. This makes PMDETA a valuable component in the synthesis and design of TRPs, which find uses in areas such as drug delivery systems, smart materials, and responsive coatings.

Application

N,N,N′,N′′,N′′ Pentamethyldiethylenetriamine can be used as:
  • A catalyst in the synthesis of multifunctional silicone acrylate prepolymers for use in UV-curable coatings.
  • A multifunctional initiating and cross-linking agent in the synthesis of polyacrylamide hydrogels. It improves the mechanical properties of the hydrogels, such as toughness and resilience, without compromising their biocompatibility.
  • An organocatalyst in ring-opening polymerization (ROP) of trimethylene carbonate. This catalyst can be easily removed after the reaction compared to metal catalysts.
  • A catalyst along with CuBr to synthesize a series of side-chain azobenzene poly(meth)acrylates via the atom transfer radical polymerization(ATRP) technique.
  • An initiator to grow polystyrene chains to prepare polyolefin-polystyrene copolymers.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Cieciórski et al.
Polymers, 13(13) (2021-07-03)
Here, we report our studies on photoresponsive poly(meth)acrylates containing azobenzene groups connected to a polymer backbone via a short methylene linker. A series of side-chain azobenzene polymers was synthesized via the atom transfer radical polymerization (ATRP) technique using several catalytic
Journal of the American Chemical Society, 112, 6190-6190 (1990)
Chun-Na Yan et al.
Polymers, 11(7) (2019-07-26)
Well-defined polymer brushes attached to nanoparticles offer an elegant opportunity for surface modification because of their excellent mechanical stability, functional versatility, high graft density as well as controllability of surface properties. This study aimed to prepare hybrid materials with good
Tae Jin Kim et al.
Polymers, 12(3) (2020-03-07)
Polyolefins (POs) are the most abundant polymers. However, synthesis of PO-based block copolymers has only rarely been achieved. We aimed to synthesize various PO-based block copolymers by coordinative chain transfer polymerization (CCTP) followed by anionic polymerization in one-pot via conversion
Jorge Royes et al.
Polymers, 11(5) (2019-05-18)
This paper describes the synthesis, thermal characterization and optical properties of liquid crystalline homopolymers and block copolymers with a repeating unit consisting of two functional units, with at least one of them being an azobenzene. Films of these polymers have

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service