Skip to Content
Merck
All Photos(1)

Key Documents

E14551

Sigma-Aldrich

Ethyl 2-bromopropionate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHBrCOOC2H5
CAS Number:
Molecular Weight:
181.03
Beilstein:
773920
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

156-160 °C (lit.)

density

1.394 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)Br

InChI

1S/C5H9BrO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3

InChI key

ARFLASKVLJTEJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengtao Lu et al.
Nature communications, 9(1), 4277-4277 (2018-10-17)
An intelligent drug release system that is triggered into action upon sensing the motion of swarmer P. mirabilis is introduced. The rational design of the drug release system focuses on a pNIPAAm-co-pAEMA copolymer that prevents drug leakage in a tobramycin-loaded
Sai Chen et al.
Polymers, 11(7) (2019-07-04)
Shape-stabilized phase change materials (SPCMs) are green, reusable energy storage materials. Because the melting temperature of n-alkyl acrylate copolymer is adjustable by controlling the side-chain length, the appropriate melting temperature can be achieved. Poly(tetradecyl acrylate-co-hexadecyl acrylate) (P(TDA-co-HDA)) with a molar
Cansu Aydogan et al.
Macromolecular rapid communications, 40(14), e1900109-e1900109 (2019-05-16)
A straightforward transformation protocol combining two distinct living polymerization methods for the controlled synthesis of block copolymers is described. In the first step, bromo-terminated poly(methyl methacrylate) is prepared by atom transfer radical polymerization (ATRP). Then, a bromide end group is
Ceren Kütahya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(46), 10444-10451 (2020-04-29)
NIR exposure at 790 nm activated photopolymerization of monomers comprising UV-absorbing moieties by using [CuII /(TPMA)]Br2 (TPMA=tris(2-pyridylmethyl)amine) in the ppm range and an alkyl bromide as initiator. Some of them comprised structural elements selected either from those showing proton transfer or
David J S Patinha et al.
Talanta, 198, 193-199 (2019-03-17)
In this work, a facile preparation of SPME fibers with increased surface area is presented. The SPME fibers were prepared by grinding poly(ionic liquids) (PILs) to obtain particles of 1-16 µm and, with the aid of a silicon adhesive, attach these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service