Skip to Content
Merck
All Photos(1)

Key Documents

54925

Sigma-Aldrich

(S)-3-Hydroxybutyric acid

≥97.0%

Synonym(s):

(+)-3-Hydroxy-n-butyric acid, (3S)-3-Hydroxybutanoic acid, (S)-β-Hydroxybutanoic acid, (S)-(+)-β-Hydroxybutyric acid, L-β-Hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720567
EC Number:
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0%

form

powder or crystals

optical activity

[α]20/D +25±1°, c = 6% in H2O

mp

45-50 °C

functional group

carboxylic acid
hydroxyl

storage temp.

2-8°C

SMILES string

C[C@H](O)CC(O)=O

InChI

1S/C4H8O3/c1-3(5)2-4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m0/s1

InChI key

WHBMMWSBFZVSSR-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-3-Hydroxybutyric acid can be used as a reagent in the synthesis of:
  • S-(2-acetamidoethyl) (S)-3-hydroxybutanethioate by reacting with acetylcysteamine in the presence of coupling reagent.[1]
  • 3-hydroxy-N-(4-methoxyphenyl)-(3S)-butanamide by treating with 4-methoxyaniline in the presence of acid-amine coupling reagent.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific cross-coupling of secondary alkyl β-trifluoroboratoamides
Sandrock DL, et al.
Journal of the American Chemical Society, 132(48), 17108-17110 (2010)
U M Nadgir et al.
Endocrine research, 27(1-2), 41-46 (2001-06-29)
We quantitated serial serum beta-hydroxybutyrate (beta-OHB) levels using the Ketosite method in 9 children with IDDM who were treated for diabetic ketoacidosis (DKA) and compared them to urinary ketone measurements by dipstick. Persistent elevations of serum beta-OHB were seen in
E Fernqvist-Forbes et al.
Diabetes care, 14(11), 1006-1012 (1991-11-01)
To study the effects of mental stress on the absorption kinetics of insulin and on glucose homeostasis and lipolysis in insulin-dependent diabetes mellitus (IDDM). Nine IDDM patients were exposed to the Stroop color word conflict test (CWT) during 40 min
Aron Altorjay et al.
World journal of gastroenterology, 11(11), 1623-1628 (2005-03-24)
With the availability of a minimally invasive approach, anti-reflux surgery has recently experienced a renaissance as a cost-effective alternative to life-long medical treatment in patients with gastroesophageal reflux disease (GERD). We are not aware of the fact whether reflux episodes
Lisa A Paige et al.
International journal of geriatric psychiatry, 22(5), 418-423 (2006-10-19)
Metabolomics, the global science of biochemistry, is an emerging field that enables detection and quantification of small molecules involved in metabolic and signaling pathways. Metabolic signatures for disease and its treatment could provide valuable biomarkers and insights about disease mechanisms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service