Skip to Content
Merck
All Photos(1)

Key Documents

54965

Sigma-Aldrich

(±)-Sodium 3-hydroxybutyrate

≥99.0% (NT)

Synonym(s):

(±)-3-Hydroxybutyric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2COONa
CAS Number:
Molecular Weight:
126.09
Beilstein:
4824866
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (NT)

mp

167-170 °C

functional group

hydroxyl

SMILES string

[Na+].CC(O)CC([O-])=O

InChI

1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

NBPUSGBJDWCHKC-UHFFFAOYSA-M

Application

(±)-Sodium 3-hydroxybutyrate has been used in a study to understand the effect of butyrate on fibroblast growth factor 21 (FGF21), which is involved in fatty acid ß-oxidation in liver. It may be used in the preparation of poly(3-hydroxybutyrate).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengying Pan et al.
Behavioural pharmacology, 31(4), 322-332 (2020-01-03)
β-hydroxybutyrate, a ketone body metabolite, has been shown to suppress depression-like behavior in rodents. In this study, we examined its antidepressive property in acute and chronic administration modes in mice by using forced swim test and tail suspension test. Results
Roman Tyzio et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(1), 34-45 (2011-01-07)
GABA depolarizes immature neurons because of a high [Cl(-)](i) and orchestrates giant depolarizing potential (GDP) generation. Zilberter and coworkers (Rheims et al., 2009; Holmgren et al., 2010) showed recently that the ketone body metabolite DL-3-hydroxybutyrate (DL-BHB) (4 mM), lactate (4
Carolina Akiko Sato Cabral Araújo et al.
PeerJ, 8, e8933-e8933 (2020-04-21)
The end of pregnancy is the period with the highest risk of occurrences of ketosis and pregnancy toxemia due to fat mobilization and increasing non-esterified fatty acids in the liver which are converted in ketone bodies, mainly β-hydroxybutyrate acid (BHB).
Sodium butyrate stimulates expression of fibroblast growth factor 21 in liver by inhibition of histone deacetylase 3.
Li H, et al.
Diabetes, 61(4), 797-806 (2012)
Novel synthesis of poly (3-hydroxybutyrate).
Juzwa M and Jedli?ski Z.
Macromolecules, 39(13), 4627-4630 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service