H6501
DL-β-Hydroxybutyric acid sodium salt
~98%, powder
Synonym(s):
(±)-3-Hydroxybutanoic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4H7O3Na
CAS Number:
Molecular Weight:
126.09
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Recommended Products
Product Name
DL-β-Hydroxybutyric acid sodium salt, ~98%
Assay
~98%
Quality Level
form
powder
solubility
water: 1 g/10 mL, clear, colorless
storage temp.
2-8°C
SMILES string
[Na].CC(O)CC(O)=O
InChI
1S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1
InChI key
NBPUSGBJDWCHKC-UHFFFAOYSA-M
Application
DL-β-Hydroxybutyric acid sodium salt has been used:
- as a standard for quantifying blood plasma and milk β-hydroxybutyric acid in Danish Jersey cows
- to investigate its anticonvulsant effects in epilepsy induced rats
- to induce hyperketonemia in Holstein dairy cows
Biochem/physiol Actions
β-Hydroxybutyric acid is a ketone body present in the rumen epithelium of liver cells. Its interconversion to acetoacetate influences the acid-base balance and is coupled NADH generation from (nicotinamide adenine dinucleotide) NAD+, measured fluorimetrically. A high plasma level of β-hydroxybutyric acid is implicated in diabetic ketoacidosis. It traverses the blood-brain barrier as a monocarboxylic acid. β-Hydroxybutyric acid may play a protective role in dopaminergic neurodegeneration in Parkinson′s disease model. β-Hydroxybutyric acid is a ligand for G protein-coupled receptor(GPR109A) and favors inhibition of secretion, transcription and translation of growth hormone (GH) and prolactin (PRL) gene in cow anterior pituitary cells (DCAPCs).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Influence of DL-beta-hydroxybutyric acid on cell proliferation and calcium influx
Cheng S, et al.
Biomacromolecules, 6(2), 593-597 (2005)
Fluorometric determination of beta-hydroxybutyrate in milk and blood plasma
Larsen T and Nielsen NI
Journal of Dairy Science, 88(6), 2004-2009 (2005)
Jianping Si et al.
Experimental and therapeutic medicine, 14(1), 765-770 (2017-07-05)
The present study aimed to investigate the anticonvulsant effects of β-hydroxybutyrate (BHB) in a kainic acid (KA)-induced rat seizure model. The concentrations of BHB and glucose were detected in the blood prior to exogenous BHB administration. Three different doses of
Jianping Si et al.
Experimental and therapeutic medicine, 20(6), 177-177 (2020-10-27)
The aim of the present study was to explore the potential anticonvulsant effects of β-hydroxybutyrate (BHB) in a kainic acid (KA)-induced rat epilepsy model. The KA-induced rat seizure model was established and BHB was administrated intraperitoneally at a dose of
Gwinyai E Chibisa et al.
Journal of animal science, 98(6) (2020-05-25)
Cattle need physically effective fiber to promote rumination and maintain rumen health, but economics favor the use of low-roughage feedlot diets. The study investigated the optimum barley silage proportion in barley-based finishing diets. Apparent total-tract digestibility (4-d total fecal collection)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service