Skip to Content
Merck
All Photos(1)

Documents

544132

Sigma-Aldrich

Bismuth neodecanoate

Synonym(s):

Bismuth trineodecanoate, Neodecanoic acid bismuth salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(OCOC(CH3)2(CH2)5CH3)3
CAS Number:
Molecular Weight:
722.75
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: bismuth
reagent type: catalyst

refractive index

n20/D 1.479 (lit.)

bp

300 °C (lit.)

density

1.145 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(C)(C)C(=O)O[Bi](OC(=O)C(C)(C)CCCCCC)OC(=O)C(C)(C)CCCCCC

InChI

1S/3C10H20O2.Bi/c3*1-4-5-6-7-8-10(2,3)9(11)12;/h3*4-8H2,1-3H3,(H,11,12);/q;;;+3/p-3

InChI key

TUQRJVHQQXIPMN-UHFFFAOYSA-K

Application

Bismuth neodecanoate can be used as a catalyst to synthesize:
  • Thermoplastic polyurethanes by reacting polycaprolactone diols, diisocyanates, and diols with anthracene group.
  • Polysiloxanes from hydroxy-terminatedpoly(dimethylsiloxanes) and ethyl silicate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV Reconfigurable Shape Memory Polyurethane with a High Recovery Ratio under Large Deformation
Hu J, et al.
Journal of Industrial and Engineering Chemistry, 60, 2144-2153 (2021)
Yiwei Kang et al.
ACS nano, 14(4), 4336-4351 (2020-04-11)
The abundant species of functional nanomaterials have attracted tremendous interests as components to construct multifunctional composites for cancer theranostics. However, their distinct chemical properties substantially require a specific strategy to integrate them in harmony. Here, we report the preparation of
Yin Chen et al.
Biomaterials science, 7(8), 3450-3459 (2019-07-04)
Bismuth-containing nanoparticles (BNPs) are potential enhancers for tumor radiotherapy. Improving the bioavailability and developing synergistic therapeutic regimens benefit the drug transformation of BNPs. In the present study, we prepare a mesoporous silica-coated bismuth nanorod (BMSNR) camouflaged by a platelet membrane
Copolymer films containing amphiphilic side chains of well-defined fluoroalkyl-segment length with biofouling-release potential
Galli G, et al.
Royal Society of Chemistry Advances, 6, 67127-67135 (2016)
Wei Hu et al.
Molecules (Basel, Switzerland), 24(23) (2019-11-30)
The high crystallization at room temperature and high cost of polyoxytetramethylene glycol (PTMG) have become obstacles to its application. To overcome these problems, a segment of PTMG can be incorporated into a block copolymer. In this work, polypropylene (PPO) glycol-polyoxytetramethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service