134481
Vinyl neodecanoate, mixture of isomers
contains 5 ppm monomethyl ether hydroquinone as inhibitor
Synonym(s):
Neodecanoic acid, vinyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C9H19CO2CH=CH2
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
contains
5 ppm monomethyl ether hydroquinone as inhibitor
refractive index
n20/D 1.436 (lit.)
bp
60-216 °C (lit.)
density
0.882 g/mL at 25 °C (lit.)
SMILES string
CCCCCCC(C)(C)C(=O)OC=C
InChI
1S/C12H22O2/c1-5-14-11(13)9-7-6-8-10-12(2,3)4/h5H,1,6-10H2,2-4H3
InChI key
TVFJAZCVMOXQRK-UHFFFAOYSA-N
Related Categories
General description
Vinyl neodecanoate, mixture of isomers is a vinyl monomer with reactive vinyl group which imparts thermal stability. Its mixture of isomers gives it a range of properties, making it flexible and stable under heat. The compound dissolves well in organic solvents, which makes it useful for coatings and drug delivery systems.
Application
Vinyl neodecanoate, mixture of isomers can be used:
- As a monomer to synthesize self-healing, super stretchable , self-adhesive homopolymers. This can be used as a matrix to fabricate semiconductive adhesives in electronic circuits and semiconducting tapes.
- As a precursor to prepare poly (vinyl acetate-co-butyl acrylate-co-neodecanoate) terpolymer matrix which acts as reaction medium and as stabilizing agent for silver nanoparticles. This can be used as antifouling coating for operation theater surfaces, medical devices.
- As a monomer to synthesize plasticizer free wood adhesive by copolymerization. It helps to enhance the water resistance of the adhesive.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
181.4 °F - closed cup
Flash Point(C)
83 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service