Skip to Content
Merck
All Photos(1)

Key Documents

542946

Sigma-Aldrich

(S)-(+)-4-Amino-3-hydroxybutyric acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +20.0°, c = 1.7 in H2O

mp

207-212 °C

functional group

amine
carboxylic acid
hydroxyl

SMILES string

NC[C@@H](O)CC(O)=O

InChI

1S/C4H9NO3/c5-2-3(6)1-4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1

InChI key

YQGDEPYYFWUPGO-VKHMYHEASA-N

General description

(S)-(+)-4-Amino-3-hydroxybutyric acid [(S)-GABOB] can be prepared starting from ethyl (S)-4-chloro-3-hydroxybutyrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Resolution of (3RS)-4-amino-3-hydroxybutanoic acid (author's transl)].
S Miyamoto et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 99(6), 642-646 (1979-06-01)
G B Melis et al.
Journal of endocrinological investigation, 5(2), 101-106 (1982-03-01)
The effect of gamma-amino beta-hydroxy butyric acid (GABOB) infusion on GH secretion has been investigated in 3 groups of 6 normal women. Different doses (100 mg/min, 20 mg/min, 3.5 mg/min) of GABOB diluted with normal saline solution were infused iv
Short synthesis of (R)-and (S)-4-amino-3-hydroxybutyric acid (GABOB).
Tiecco M, et al.
Synthesis, 2005(04), 579-582 (2005)
Different efficacies of d- and l-gamma-amino-beta-hydroxybutyric acids in GABA receptor and transport test systems.
E Roberts et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 1(2), 132-140 (1981-02-01)
S Banfi et al.
Il Farmaco; edizione scientifica, 39(1), 16-22 (1984-01-01)
A number of N-substituted 4-hydroxy-, 4-acyloxy- and 4-alkoxy-2-pyrrolidinones were examined by a screening method predictive of their activity on cognitive processes. The 1-aminocarbonylmethyl-substituted compounds showed a favorable effect on learning and memory, and among them the most active was the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service