Merck
  • Chitin-glucan complex production by Komagataella pastoris: Downstream optimization and product characterization.
Chitin-glucan complex production by Komagataella pastoris: Downstream optimization and product characterization.
Carbohydrate polymers (2015-06-17)
Inês Farinha, Paulo Duarte, Ana Pimentel, Evgeniya Plotnikova, Bárbara Chagas, Luís Mafra, Christian Grandfils, Filomena Freitas, Elvira Fortunato, Maria A M Reis
ABSTRACT
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic salts contents (3.0 and 0.9 wt%, respectively). CGCpure had an average molecular weight of 4.9 × 10(5)Da with a polydispersity index of 1.7, and a crystallinity index of 50%. Solid-state NMR provided structural insight at the co-polymer. X-ray diffraction suggests that CGCpure has α-chitin in its structure. CGCpure presented an endothermic decomposition peak at 315°C, assigned to the degradation of the saccharide structures. This study revealed that K. pastoris CGC has properties similar to other chitinous biopolymers and may represent an attractive alternative to crustacean chitin derived-products, being a reliable raw material for the development of new/improved pharmaceutical, cosmetic or food products.
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ special grade
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ first grade, ≥99.0%
SAFC
N,N-Dimethylacetamide, Ph. Eur.