Skip to Content
Merck
All Photos(1)

Key Documents

511382

Sigma-Aldrich

1-Boc-hexahydro-1,4-diazepine

98%

Synonym(s):

1-Boc-homopiperazine, tert-Butyl homopiperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
Beilstein:
7581789
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.471 (lit.)

bp

95-110 °C/0.5 mmHg (lit.)

density

1.016 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCCNCC1

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-7-4-5-11-6-8-12/h11H,4-8H2,1-3H3

InChI key

WDPWEXWMQDRXAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Boc-hexahydro-1,4-diazepine is an N-boc protected homopiperazine. Its enthalpy of vaporization at boiling point (558.15K) is 49.498kjoule/mol.

Application

Used in synthesis of potent anticoagulant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Koshio et al.
Bioorganic & medicinal chemistry, 12(9), 2179-2191 (2004-04-15)
Factor Xa (fXa) is a serine protease involved in the coagulation cascade, which has received great interest as a potential target for the development of new antithrombotic drugs. Herein we report a novel series of fXa inhibitors in which the
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 586-586 (2014)

Articles

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service