Skip to Content
Merck
All Photos(3)

Key Documents

H10401

Sigma-Aldrich

Hexamethyleneimine

99%

Synonym(s):

1-Azacycloheptane, Azepane, HMI, Perhydroazepine, Hexahydro-1H-azepine, Homopiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein:
1084
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

7.4 mmHg ( 21.1 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

138 °C/749 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

C1CCCNCC1

InChI

1S/C6H13N/c1-2-4-6-7-5-3-1/h7H,1-6H2

InChI key

ZSIQJIWKELUFRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexamethyleneimine (HMI) can be used in the preparation of:
  • zeolitic catalysts such as MCM-22 and MCM-49[1][2][3]
  • SAPO-35, a molecular sieve[4][5]
  • various microporous aluminosilicates and titanosilicates[6][7][8]
  • organic-inorganic hybrid pervoskite[9]


It can also been used in the total synthesis of various natural products and bioactive molecules such as (−)-balanol, indole estrogens, derivatives of daunorubicin and (+)-cetiedil.[10][11][12][13]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel titanosilicate with MWW structure. I. Hydrothermal synthesis, elimination of extra framework titanium, and characterizations.
Wu P, et al.
The Journal of Physical Chemistry B, 105(15), 2897-2905 (2001)
Formal total synthesis of (-)-balanol: Concise approach to the hexahydroazepine segment based on RCM.
Furstner A and Thiel OR
The Journal of Organic Chemistry, 65(6), 1738-1742 (2000)
J R Lundkvist et al.
Journal of medicinal chemistry, 33(12), 3182-3189 (1990-12-01)
A series of conformationally restricted analogues of the partial muscarinic agonist N-methyl-N-(1-methyl-4-pyrrolidino-2-butynyl)acetamide (BM 5; 1) was synthesized. Three of the racemic derivatives were resolved into the enantiomers. The compounds were investigated for muscarinic and antimuscarinic activity in the isolated guinea
SAPO-35 Molecular Sieve: Synthesis, Characterization, and Adsorbate Interactions of Cu (II) in CuH? SAPO-35.
Prakash AM, et al.
Chemistry of Materials, 10(3), 932-941 (1998)
Efficiency of zeolite MCM-22 with different SiO2/Al2O3 molar ratios in gas phase glycerol dehydration to acrolein.
Carrico CS, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 181, 74-82 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service