Skip to Content
Merck
All Photos(2)

Key Documents

488216

Sigma-Aldrich

3,4-Dihydroxy-1-butene

≥99%

Synonym(s):

3-Butene-1,2-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH(OH)CH2OH
CAS Number:
Molecular Weight:
88.11
Beilstein:
1633578
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

bp

195 °C/733 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC(O)C=C

InChI

1S/C4H8O2/c1-2-4(6)3-5/h2,4-6H,1,3H2

InChI key

ITMIAZBRRZANGB-UHFFFAOYSA-N

General description

3,4-Dihydroxy-1-butene, also known as 3-butene-1,2-diol (BDdiol), is a metabolite of 1,3-butadiene. It forms the precursor for synthesizing different chiral building blocks. BDdiol can undergo oxidation to form hydroxymethylvinyl ketone (HMVK). 1,2-epoxy-3-butene (EB) on hydrolysis in the presence of epoxide hydrolases (EH) forms BDdiol.

Application

3,4-Dihydroxy-1-butene can be used:
  • As a reactant to synthesize cyclic organic carbonates by continuous flow procedure.
  • To prepare substituted oxazolidinone ligands used to target medicinally relevant RNAs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-Butene-1, 2-diol: An attractive precursor for the synthesis of enantiomerically pure organic compounds.
Rao AVR, et al.
Tetrahedron, 45(22), 7031-7040 (1989)
Versatile and scalable synthesis of cyclic organic carbonates under organocatalytic continuous flow conditions
Gerardy R, et al.
Catalysis Science & Technology, 9(24), 6841-6851 (2019)
Gregory M Sandala et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(19), 4865-4873 (2009-03-28)
High-level conventional ab initio and density functional theory (DFT) calculations have been performed to examine the fate of the native substrate glycerol (1) and its analogue but-3-ene-1,2-diol (7) in the coenzyme B(12)-dependent enzyme glycerol dehydratase (GDH). Experimental studies find that
X Cheng et al.
Drug metabolism and disposition: the biological fate of chemicals, 21(1), 121-124 (1993-01-01)
A rapid, simple extraction and GC assay procedure is described that allows quantitation of micromolar concentrations of butadiene bisoxide and 3-butene-1,2-diol in microsomal suspensions exposed to butadiene. Butane-1,4-diol is used as the internal standard. The recovery of these compounds from
R J Krause et al.
Chemical research in toxicology, 14(12), 1590-1595 (2001-12-18)
The metabolic fate of 3-butene-1,2-diol (BDD), a secondary metabolite of the industrial carcinogen, 1,3-butadiene, is unclear. The current study characterizes BDD oxidation to hydroxymethylvinyl ketone (HMVK), a reactive Michael acceptor. Because of its instability in aqueous medium, HMVK was trapped

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service