Skip to Content
Merck
All Photos(1)

Key Documents

52490

Sigma-Aldrich

2,4-Hexadiyne-1,6-diol

≥98.0% (GC)

Synonym(s):

1,6-Dihydroxy-2,4-hexadiyne, 2,4-Hexadiynediol, Diacetylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C≡CC≡CCH2OH
CAS Number:
Molecular Weight:
110.11
Beilstein:
773791
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

mp

113-114 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

OCC#CC#CCO

InChI

1S/C6H6O2/c7-5-3-1-2-4-6-8/h7-8H,5-6H2

InChI key

JXMQYKBAZRDVTC-UHFFFAOYSA-N

General description

2,4-Hexadiyne-1,6-diol can be prepared from propargyl alcohol. 2,4-Hexadiyne-1,6-diol readily undergoes polymerization when heated under vacuum or inert gas atmosphere.

Application

2,4-Hexadiyne-1,6-diol may be used as a starting material in the synthesis of thiarubrine A (an antibiotic). It may also be used to synthesize disodium salt of 2,4-hexadiyne 1,6-disulfate (HDDS).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The low-temperature polarized optical absorption of a crystalline diacetylene: 2, 4-hexadiyne-1, 6-diol.
Kawaoka, K.
Chemical Physics Letters, 37(3), 561-565 (1976)
Self-assembled alternating multilayers built-up from diacetylene bolaamphiphiles and poly (allylamine hydrochloride): polymerization properties, structure, and morphology.
Saremi F, et al.
Langmuir, 11(4), 1068-1071 (1995)
Solid-state thermal polymerization of 2, 4-hexadiyne-1, 6-diol.
Bloor D and Stevens GC.
Journal of Polymer Science. Part B, Polymer Physics, 15(4), 703-714 (1977)
Notes-use of amines in the glaser coupling reaction.
Cameron M and Bennett G.
The Journal of Organic Chemistry, 22(5), 557-558 (1957)
Chemistry of 1, 2-dithiins. Synthesis of the potent antibiotic thiarubrine A.
Koreeda M and Yang W.
Journal of the American Chemical Society, 116(23), 10793-10794 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service