Skip to Content
Merck
All Photos(3)

Documents

496820

Sigma-Aldrich

4-Vinyl-1,3-dioxolan-2-one

99%

Synonym(s):

VEC, Vinyl ethylene carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.45 (lit.)

bp

237 °C/733 mmHg (lit.)

density

1.188 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1COC(=O)O1

InChI

1S/C5H6O3/c1-2-4-3-7-5(6)8-4/h2,4H,1,3H2

InChI key

BJWMSGRKJIOCNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinyl-1,3-dioxolan-2-one (VEC) is a cyclic carbonate, that can be synthesized from acrolein by reacting with sulfur ylide and CO2. The ruthenium-catalyzed transfer hydrogenation (TH) of VEC in the presence of 2-propanol forms 1,2-butanediol. Indolines and indoles undergo selective C-H allylation with VEC in the presence of rhodium(III) catalyst.

Application

4-Vinyl-1,3-dioxolan-2-one may be used in the preparation of 2-arylbenzo[d]thiazole scaffolds.
It may be used in the synthesis of the following multi-functional cyclic carbonates:
  • 4,4′-((hexane-1,6-diylbis(sulfanediyl))bis-(ethane-2,1-diyl))bis(1,3-dioxolan-2-one) (Bis-CC)
  • 2-ethyl-2-(((3-((2-(2-oxo-1,3-dioxolan-4-yl)-ethyl)thio)propanoyl)oxy) methyl)propane-1,3-diyl bis(3-((2-(2-oxo-1,3-dioxolan-4-yl)ethyl)thio)propanoate) (Tris-CC)
  • 2,2-bis(((3-((2-(2-oxo-1,3-dioxolan-4-yl)ethyl)-thio)propanoyl)oxy) methyl)propane-1,3-diyl bis(3-((2-(2-oxo-1,3-dioxolan-4-yl)ethyl)thio)propanoate) (Tetra-CC)
Used to synthesize functional polymers.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

206.4 °F - closed cup

Flash Point(C)

96.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

William A Rose et al.
PloS one, 7(3), e32728-e32728 (2012-03-14)
The human vaginal microbiome plays a critical but poorly defined role in reproductive health. Vaginal microbiome alterations are associated with increased susceptibility to sexually-transmitted infections (STI) possibly due to related changes in innate defense responses from epithelial cells. Study of
Rhodium(III)-catalyzed heteroatom-directed C-H allylation with allylic phosphonates and allylic carbonates at room temperature.
Jo H, et al.
Tetrahedron, 72(4), 571-578 (2016)
Giovanna Berruti et al.
Histochemistry and cell biology, 150(2), 173-185 (2018-05-26)
Spermatids are haploid differentiating cells that, in the meantime they differentiate, translocate along the seminiferous epithelium towards the tubule lumen to be just released as spermatozoa. The success of such a migration depends on dynamic of spermatid-Sertoli cell contacts, the
John R Brannon et al.
Nature communications, 11(1), 2803-2803 (2020-06-06)
Host-associated reservoirs account for the majority of recurrent and oftentimes recalcitrant infections. Previous studies established that uropathogenic E. coli - the primary cause of urinary tract infections (UTIs) - can adhere to vaginal epithelial cells preceding UTI. Here, we demonstrate
Progress in Organic Coatings, 47, 77-86 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service