Skip to Content
Merck
All Photos(2)

Documents

483095

Sigma-Aldrich

PEDOT:PSS

greener alternative

conductive grade, 1.3 wt. % aqueous dispersion

Synonym(s):

PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate), 1.3 wt % dispersion in H2O, conductive grade

grade

conductive grade

composition

PEDOT content, 0.5 wt. %
PSS content, 0.8 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.3 wt % dispersion in H2O

band gap

1.6 eV

conductivity

1 S/cm

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

A conducting polymer such as poly(3,4-ethylenedioxythiophene) doped with poly(styrene sulfonate) anions (PEDOT/PSS) is widely used in various organic optoelectronic devices. High electrical conductivity and good oxidation resistance of such polymers make it suitable for electromagnetic shielding and noise suppression. Thus, the polymer film was found to possess high transparency throughout the visible light spectrum and even into near IR and near UV regions, virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend. Impact of small electric and magnetic fields on the polymer was studied.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is used in energy conversion and storage, thus has been enhanced for energy efficiency. Click here for more information.

Application

PEDOT:PSS polymeric films have been used as a charge dissipation layer in electron-beam lithography and focused ion beam milling. Sample preparation has been reported to be easier and quicker for various substrates, including gallium nitride (GaN) on sapphire (Al2O3) substrates, zinc oxide (ZnO), fused silica, lithium niobate (LiNbO3), silicon carbide (SiC) and diamond (C), spin-coated onto the ITO coated glass substrate. PEDOT: PSS layers have also been reported to be used as anode buffer layer for organic solar cells and as replacements for the transparent conductive coatings of organic solar cells. Various studies report the use of metal modified conductive grade PEDOT: PSS as an anode buffer layer in solar cells, example: copper phthalocyanine/fullerene-based solar cells 4 Conductive PEDOT:PSS combined with polyvinylidene fluoride (PVDF) membranes may be used to prepare PEDOT:PSS-PVDF ionic liquid soft actuators. The function of PEDOT:PSS as a pseudocapacitive material was investigated.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Features and Benefits

Antistat coating for plastic and glass.

Packaging

Packaged in glass bottles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faramarz Hossein-Babaei et al.
Scientific reports, 7, 42299-42299 (2017-02-17)
Many gaseous markers of critical biological, physicochemical, or industrial occurrences are masked by the cross-sensitivity of the sensors to the other active components present at higher concentrations. Here, we report the strongly selective diffusion and drift of contaminant molecules in
Yi-Kuang Yen et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-12)
Detecting the concentration of Pb2+ ions is important for monitoring the quality of water due to it can become a health threat as being in certain level. In this study, we report a nanomechanical Pb2+ sensor by employing the complementary
Conductive Polymers for Advanced Micro- and Nano-fabrication Processes
<B>Dylewicz R, et al.</B>
Material Matters, 6(1) null
PEDOT: PSS layers as replacements for the transparent conductive coatings of organic solar cells.
Schwertheim S, et al.
Photovoltaic Specialists Conference, 1991., Conference Record of the Twenty Second IEEE (2010)
Zisheng Su et al.
Nanoscale research letters, 7(1), 465-465 (2012-08-21)
Ultraviolet-ozone-treated poly(3,4-ethylenedioxythiophene):poly(styrene sulfonate) (PEDOT:PSS)was used as the anode buffer layer in copper phthalocyanine (CuPc)/fullerene-based solar cells. The power conversion efficiency of the cells with appropriated UV-ozone treatment was found to increase about 20% compared to the reference cell. The improved

Articles

Sigma-Aldrich tutorial on lithography nanopatterning explains patterning layers for conductors, semiconductors, and dielectrics on surfaces.

Sigma-Aldrich tutorial on lithography nanopatterning explains patterning layers for conductors, semiconductors, and dielectrics on surfaces.

Sigma-Aldrich tutorial on lithography nanopatterning explains patterning layers for conductors, semiconductors, and dielectrics on surfaces.

Sigma-Aldrich tutorial on lithography nanopatterning explains patterning layers for conductors, semiconductors, and dielectrics on surfaces.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service