Skip to Content
Merck
All Photos(3)

Key Documents

483028

Sigma-Aldrich

3,4-Ethylenedioxythiophene

97%

Synonym(s):

2,3-Dihydrothieno[3,4-b]-1,4-dioxin, EDOT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2S
CAS Number:
Molecular Weight:
142.18
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.5765 (lit.)

bp

193 °C (lit.)

density

1.331 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1COc2cscc2O1

InChI

1S/C6H6O2S/c1-2-8-6-4-9-3-5(6)7-1/h3-4H,1-2H2

InChI key

GKWLILHTTGWKLQ-UHFFFAOYSA-N

General description

3,4-Ethylenedioxythiophene (EDOT) is an electro-active conductive monomer with a thiol group that combines an electron donor and electron acceptor in a donor-acceptor-donor arrangement.[1]
Monomer used in the synthesis of conducting polymers.

Application

EDOT can be polymerized to form poly(3,4-ethylenedioxythiophene) (PEDOT) for use as an electrochromic polymer (EC) based coating for a variety of applications like solid state organic electrochemical supercapacitors (OESCs),[2][3] electrochromic devices (ECDs),[1] and carbon nanotubes (CNTs) based electrochemical devices for diabetes monitoring.[4]
It can be used:
  • As a reductant in a one-pot synthesis of gold nanoparticles from HAuCl4 (254169).[5]
  • As a starting material used in palladium-catalyzed mono- and bis-arylation reactions.[6]
  • In the synthesis of conjugated polymers and copolymers, with potential optical applications.[7][8]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecules, 37, 4087-4098 (2004)
Protein-mediated synthesis of the conducting polymer PEDOT: PSS.
Hira SM and Payne CK
Synthetic Metals, 176(5), 104-107 (2013)
Towards sustainable solid-state supercapacitors: electroactive conducting polymers combined with biohydrogels.
Perez-Madrigal MM, et al.
Journal of Material Chemistry A, 4(5), 1792-1805 (2016)
Shanmugam Senthil Kumar et al.
Langmuir : the ACS journal of surfaces and colloids, 23(6), 3401-3408 (2007-02-08)
Herein, we report a one-pot synthesis of highly stable Au nanoparticles (AuNPs) using 3,4-ethylenedioxythiophene (EDOT) as a reductant and polystyrene sulfonate (PSS-) as a dopant for PEDOT and particle stabilizer. The synthesis demonstrated in this work entails the reduction of
Yuzhe Xie et al.
Biosensors & bioelectronics, 87, 31-37 (2016-08-16)
A motor-based microprobe is proposed using a tubular microengine powered by bio-assembled enzyme as catalyst and exploited for washing-free detection of DNA through motion readout. The microprobe is fabricated by assembling a catalase layer on the inner surface of poly(3,4-ethylenedioxythiophene)/Au

Articles

Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service