Skip to Content
Merck
All Photos(1)

Documents

477923

Sigma-Aldrich

9H-Fluorene-9,9-dimethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

142-145 °C (lit.)

SMILES string

OCC1(CO)c2ccccc2-c3ccccc13

InChI

1S/C15H14O2/c16-9-15(10-17)13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,16-17H,9-10H2

InChI key

RHBLISBUFROBBC-UHFFFAOYSA-N

General description

9H-Fluorene-9,9-dimethanol is an aryldiol.

Application

9H-Fluorene-9,9-dimethanol may be used in the synthesis of:
  • 4,4′-(9H-fluorene-9-yl-dimethoxy)diphthalonitrile]
  • poly- [oxy(9H-fluoren-9,9-dimethylene)oxy(tetramethyldisilylene)]
  • 9,9-(Ph2POCH2)2-fluorene, a diphosphinite ligand

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, characterization, electrochemistry and VOC sensing properties of novel ball-type dinuclear metallophthalocyanines.
Cimen Y, et al.
Sensors and Actuators B, Chemical, 202, 1137-1147 (2014)
A fluorene-based diphosphinite ligand, its Ni, Pd, Pt, Fe, Co and Zn complexes and the first structurally characterized diphosphinate metal chelates.
Li C, et al.
Inorgorganica Chimica Acta, 363(15), 4337-4345 (2010)
Synthesis and Properties of Poly [oxy (arylene) oxy (tetramethyldisilylene)] s via Melt Copolymerization Reaction.
Jung EA and Park YT.
Bull. Korean Chem. Soc., 34(6), 1637-1642 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service