Skip to Content
Merck
All Photos(3)

Key Documents

H31204

Sigma-Aldrich

9-Hydroxyfluorene

96%

Synonym(s):

9-Fluorenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

153-154 °C (lit.)

SMILES string

OC1c2ccccc2-c3ccccc13

InChI

1S/C13H10O/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8,13-14H

InChI key

AFMVESZOYKHDBJ-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Chen et al.
Drug metabolism and disposition: the biological fate of chemicals, 12(4), 421-426 (1984-07-01)
Fluoren-9-ol and fluoren-9-one were used as model substrates to study microsomal metabolism of alcohols and carbonyl compounds. It was found that there was an oxidoreductase(s) present in the microsomal preparation that catalyzed interconversion of this alcohol and ketone using pyridine
L Bastiaens et al.
Research in microbiology, 152(10), 849-859 (2002-01-05)
The promoter probe mini-Tn5-luxAB-tet was used to create a luxAB transcriptional fusion responding to fluorene in the fluorene utilising bacterium Sphingomonas sp. LB126. The mutant strain, named L-132, was impaired in fluorene utilisation and strongly emitted light upon addition of
Biban Gill et al.
Analytical and bioanalytical chemistry, 411(7), 1397-1407 (2019-01-27)
Urinary 1-hydroxypyrene (OH-Pyr) is widely used for biomonitoring human exposures to polycyclic aromatic hydrocarbons (PAHs) from air pollution and tobacco smoke. However, there have been few rigorous validation studies reported to ensure reliable OH-Pyr determination for occupational health and risk
Pooja V Reddy et al.
Archives of microbiology, 200(1), 171-182 (2017-09-25)
A high-efficiency fluoranthene-degrading bacterium Paenibacillus sp. PRNK-6 was isolated from PAH-contaminated soil. The strain degrades 96% (240 mg l
Microbial reduction of 9-fluorenone to 9-hydroxyfluorene in carbon-filtered water: a confounding factor in an aquatic bioassay of 9-fluorenone with larvae of the midge, Chironomus tentans (Fabr.).
P W O'Keefe et al.
Bulletin of environmental contamination and toxicology, 75(6), 1060-1066 (2006-01-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service