Skip to Content
Merck
All Photos(1)

Key Documents

757187

Sigma-Aldrich

2,7-Dibromo-9,9-dimethyl-9H-fluorene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12Br2
CAS Number:
Molecular Weight:
352.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

177-181 °C

SMILES string

CC1(C)c2cc(Br)ccc2-c3ccc(Br)cc13

InChI

1S/C15H12Br2/c1-15(2)13-7-9(16)3-5-11(13)12-6-4-10(17)8-14(12)15/h3-8H,1-2H3

InChI key

LONBOJIXBFUBKQ-UHFFFAOYSA-N

Application

This material is a precursor to a number of organic semiconducting polymers for OPV as well as a variety of hole transport for OLED devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service