Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene glycol) diacrylate
Synonym(s):
PEG diacrylate
Linear Formula:
C
3
H
3
O(OCH
2
CH
2
)
n
C
3
H
3
O
2
CAS Number:
26570-48-9
Products
Brand
Sigma-Aldrich (9)
Color
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
455008
Poly(ethylene glycol) diacrylate,
average M
n
700, acrylate, 100 ppm MEHQ as inhibitor, 300 ppm BHT as inhibitor
Expand
View Pricing
475629
Poly(ethylene glycol) diacrylate,
average M
n
250, acrylate, 100 ppm MEHQ as inhibitor
Expand
View Pricing
437441
Poly(ethylene glycol) diacrylate,
average M
N
575, cross-linking reagent polymerization reactions, acrylate, 400-600 ppm MEHQ as inhibitor
Expand
View Pricing
701963
Poly(ethylene glycol) diacrylate,
average M
n
6,000, acrylate, ≤1,500 ppm MEHQ as inhibitor
Expand
View Pricing
729086
Poly(ethylene glycol) diacrylate,
average M
n
1,000, acrylate, MEHQ as inhibitor (may contain)
Expand
View Pricing
729094
Poly(ethylene glycol) diacrylate,
average M
n
10,000, acrylate, MEHQ as inhibitor (may contain)
Expand
View Pricing
907227
Poly(ethylene glycol) diacrylate,
average M
n
4,000, average M
n
4,000, <200 ppm MEHQ as inhibitor
Expand
View Pricing
929751
Poly(ethylene glycol) diacrylate,
average M
N
3,400, acrylate, MEHQ as inhibitor
Expand
View Pricing
701971
Poly(ethylene glycol) diacrylate,
average M
n
2,000, acrylate, MEHQ as inhibitor (may contain)
Expand
View Pricing