Skip to Content
Merck
All Photos(1)

Documents

454990

Sigma-Aldrich

Poly(ethylene glycol) methyl ether acrylate

average Mn 480, contains 100 ppm MEHQ as inhibitor, 100 ppm BHT as inhibitor

Synonym(s):

Acryl-PEG, Methoxy PEG acrylate, Methoxy poly(ethylene glycol) monoacrylate, Poly(ethylene glycol) monomethyl ether monoacrylate, mPEG-acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn 480

contains

100 ppm BHT as inhibitor
100 ppm MEHQ as inhibitor

density

1.09 g/mL at 25 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) methyl ether methacrylate (PEGMA) is a nonlinear analog of polyethylene glycol (PEG). It is a biocompatible homopolymer with a brush type structure that is mainly used to provide a PEG modified surface.

Application

PEGMA can be used in the development of molecular brushes as a substrate material for stimuli-responsive surfaces in biomedical applications. It may be used in the surface modification of poly(ether sulfone) based ultrafiltration (UF) membrane as a foul-resistant material.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new precursor for conducting polymer-based brush interfaces with electroactivity in aqueous solution.
Strover LT, et al.
Polymer, 54(4), 1305-1317 (2013)
Using co-solvents and high throughput to maximize protein resistance for poly (ethylene glycol)-grafted poly (ether sulfone) UF membranes
Yune PS, et al.
Journal of Membrane Science , 370(1-2), 166-174 (2011)
Surface-initiated photopolymerization of poly (ethylene glycol) methyl ether methacrylate on a diethyldithiocarbamate-mediated polymer substrate
Luo Ning, et al.
Macromolecules, 35(7), 2487-2493 (2002)
Self-assembly of brush-like poly [poly (ethylene glycol) methyl ether methacrylate] synthesized via aqueous atom transfer radical polymerization
Hussain H, et al.
Langmuir, 24(23), 13279-13286 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service