Skip to Content
Merck
All Photos(2)

Key Documents

457698

Sigma-Aldrich

(R)-(+)-2-Methyl-CBS-oxazaborolidine solution

1 M in toluene

Synonym(s):

α,α-Diphenyl-D-prolinolmethylboronic acid cyclamide ester, (R)-1-Methyl,3,3-diphenyl-tetrahydro-pyrrolo(1,2-c)(1,3,2)oxazaborole, (R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

1 M in toluene

bp

111 °C

density

0.95 g/mL at 25 °C

functional group

phenyl

storage temp.

room temp

SMILES string

[H][C@]12CCCN1B(C)OC2(c3ccccc3)c4ccccc4

InChI

1S/C18H20BNO/c1-19-20-14-8-13-17(20)18(21-19,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17H,8,13-14H2,1H3/t17-/m1/s1

InChI key

VMKAFJQFKBASMU-QGZVFWFLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-2-Methyl-CBS-oxazaborolidine solution [1M in toluene] may be used as a catalyst in the asymmetric borane reduction of perfluoroalkyl ketones.
It may also be used in the preparation of:
  • (-)-diospongin B
  • (1R)-2-azido-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol
  • (S)-α-deuteriobenzyl alcohol
  • (3S,4R,5S)-1-(trimethylsilyl)-4,5-epoxyhex-1-yn-3-ol
Excellent catalyst of asymmetric reductions.
The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols.
Used in a desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.

Physical form

Precipitate may form in storage, but does not affect product quality. Gently heat product to 80−90 °C under an inert atmosphere to redissolve solids.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomeric resolution of fluorous mixture by chiral CD columns: asymmetric reduction of a mixture of fluorous ketones.
Nakamura Y, et al.
Tetrahedron Letters, 44(33), 6221-6225 (2003)
The discovery of long-acting saligenin ? 2 adrenergic receptor agonists incorporating hydantoin or uracil rings.
Procopiou PA, et al.
Bioorganic & Medicinal Chemistry, 19(14), 4192-4201 (2011)
Stereoselective glycosylations of a family of 6-deoxy-1, 2-glycals generated by catalytic alkynol cycloisomerization.
McDonald FE, et al.
Journal of the American Chemical Society, 122(18), 4304-4309 (2000)
Bis (oxazoline)-ligand-mediated asymmetric [2, 3]-Wittig rearrangement of benzyl ethers: reaction mechanism based on the hydrogen/deuterium exchange effect.
Kitamura M, et al.
Tetrahedron, 68(22), 4280-4285 (2012)
Singh, V. K.
Synthesis, 605-605 (1992)

Articles

we are pleased to offer both enatiomers of 2-methyl-CBS-oxazaborolidine as a dry reagent, in addition to our current offerings as a 1M solution in toluene.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service