The black CTFE body or cylinder portion of this electrode has a diameter of 6.4 mm.
BASMW1032
Platinum Wire Auxiliary Electrode
Synonym(s):
Auxiliary Pt electrode
About This Item
Recommended Products
material
CTFE cylinder
gold connector (plated)
electrode L
7.5 cm
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the diameter of the CTFE cylinder part
1 answer-
Helpful?
-
-
Hello, I have doubts about the size of the electrode. 7.5 cm means the whole size of the electrode, or Pt wire only? Actually, we are using Pt Wire Electrode (0.5x50), but I want to buy more than that size.
1 answer-
According to the manufacturer BASi Research Products, this product is a 7.5 cm long platinum wire (0.5 mm diameter) auxiliary/counter electrode.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service