Skip to Content
Merck
All Photos(1)

Key Documents

128910

Sigma-Aldrich

N,O-Bis(trimethylsilyl)acetamide

synthesis grade, ≥95%

Synonym(s):

BSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
203.43
Beilstein:
1306669
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

synthesis grade

Assay

≥95%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

71-73 °C/35 mmHg (lit.)

density

0.832 g/mL at 20 °C (lit.)

SMILES string

C\C(O[Si](C)(C)C)=N/[Si](C)(C)C

InChI

1S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3/b9-8+

InChI key

SIOVKLKJSOKLIF-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)acetamide(BSA) is a powerful silylating agent for the protection of amides, amines, alcohols, carboxylic acids, enols and phenols. It can also be used as a Bronsted base precursor in Tsuji–Trost reactions. Additionally, it can be used to activate different functional groups during the production of nucleosides, peptides, and heterocycles.

Application

Derivatization reagent for GC-MS analysis of phenolic acids in fruits.
Reagent for the regioselective desulfation of polysaccharide sulfates. Also used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

Features and Benefits

BSA is good moisture absorbent.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solvent free synthesis of 3, 4-dihydropyrimidine-2-(1H)-ones/thiones catalyzed by N, O-bis (trimethylsilyl) acetamide and dicyclohexyl carbodimide
Murthy YLN, et al.
Arabian Journal of Chemistry, 9, S1740-S1746 (2016)
Carbohydrate Research, 237, 313-313 (1992)
Mohammad Saraji et al.
Journal of separation science, 29(9), 1223-1229 (2006-07-13)
A simple analytical procedure based on single-drop microextraction combined with in-syringe derivatization and GC-MS was developed for determination of some phenolic acids in fruits and fruit juices. Cinnamic acid, o-coumaric acid, caffeic acid, and p-hydroxybenzoic acid were used as model
N, O-Bis (trimethylsilyl) acetamide
Claraz A
Synlett, 24(5), 657-658 (2013)
Olga Moiseeva et al.
Cell cycle (Georgetown, Tex.), 14(15), 2408-2421 (2015-06-02)
Expression of oncogenes or short telomeres can trigger an anticancer response known as cellular senescence activating the p53 and RB tumor suppressor pathways. This mechanism is switched off in most tumor cells by mutations in p53 and RB signaling pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service