441171
(Bromomethyl)cyclobutane
97%
Synonym(s):
Cyclobutylmethyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4H7CH2Br
CAS Number:
Molecular Weight:
149.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.48 (lit.)
bp
123-124 °C (lit.)
density
1.326 g/mL at 25 °C (lit.)
functional group
bromo
SMILES string
BrCC1CCC1
InChI
1S/C5H9Br/c6-4-5-2-1-3-5/h5H,1-4H2
InChI key
FLHFTXCMKFVKRP-UHFFFAOYSA-N
Related Categories
General description
(Bromomethyl)cyclobutane (bromomethyl-cyclobutane) is an aryl fluorinated building block. Its reaction with various hydroxychromen-4-one derivatives in the presence of anhydrous K2CO3/dry acetone and Bu4N+I- (PTC) has been investigated.
Application
(Bromomethyl)cyclobutane may be used in the synthesis of racemic 3-cyclobutylalanine and 17-(cyclobutylmethyl)morphinan-3-ol (butorphan).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
105.8 °F - closed cup
Flash Point(C)
41 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Photochemical synthesis and antimicrobial studies of new chromen-4-one based vinyl ethers.
Yusuf M, et al.
Arabian Journal of Chemistry (2013)
Markus Nett et al.
Journal of medicinal chemistry, 52(19), 6163-6167 (2009-09-15)
The natural proteasome inhibitor salinosporamide A from the marine bacterium Salinispora tropica is a promising drug candidate for the treatment of multiple myeloma and mantle cell lymphoma. Using a comprehensive approach that combined chemical synthesis with metabolic engineering, we generated
Michael Decker et al.
Journal of medicinal chemistry, 53(1), 402-418 (2009-11-26)
The phenolic group of the potent mu and kappa opioid morphinan agonist/antagonists cyclorphan and butorphan was replaced by phenylamino and benzylamino groups including compounds with para-substituents in the benzene ring. These compounds are highly potent mu and kappa ligands, e.g.
Ao Zhang et al.
Journal of medicinal chemistry, 47(1), 165-174 (2003-12-30)
A series of 10-ketomorphinan analogues were synthesized, and their binding affinity at all three opioid receptors was investigated. In most cases, high affinity at micro and kappa receptors, and lower affinity at delta receptor was observed, resulting in good selectivity
Cyclopropylmethyl-and cyclobutylmethyllithium by an arene-catalyzed lithiation. Stability and reactivity.
Pe?afiel I, et al.
Tetrahedron, 66(16), 2928-2935 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service