Skip to Content
Merck
All Photos(1)

Documents

713457

Sigma-Aldrich

(Iodomethyl)cyclopropane

≥95.0% (GC)

Synonym(s):

Cyclopropylmethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CH2I
CAS Number:
Molecular Weight:
182.00
Beilstein:
1900420
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.542

SMILES string

ICC1CC1

InChI

1S/C4H7I/c5-3-4-1-2-4/h4H,1-3H2

InChI key

JHKJQWFHNIOUKY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service