Skip to Content
Merck
All Photos(2)

Key Documents

437883

Sigma-Aldrich

Strontium acetate

99.95% trace metals basis

Synonym(s):

Strontium diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)2Sr
CAS Number:
Molecular Weight:
205.71
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

powder

reaction suitability

core: strontium

impurities

~4% water

SMILES string

CC(=O)O[Sr]OC(C)=O

InChI

1S/2C2H4O2.Sr/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

RXSHXLOMRZJCLB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to promising candidate materials for superconductor wire.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee, B.I. et al.
Physica C, 226, 377-377 (1994)
N C A Claydon et al.
The Journal of clinical dentistry, 20(5), 158-166 (2009-11-12)
The aim of these clinical studies was to evaluate an in situ dentine tubule occlusion model, and to determine the occluding effect from novel occluding agents on patent dentine tubules compared to a positive control (8% strontium acetate--Sensodyne Mint) and
N X West et al.
Journal of clinical periodontology, 24(4), 209-215 (1997-04-01)
A considerable number of varied agents are apparently effective in the treatment of dentine hypersensitivity. In particular, the literature supports the efficacy of strontium, potassium and fluoride containing toothpastes. This study was a double-blind, randomised, parallel group comparison of three
R Patel et al.
The Journal of clinical dentistry, 22(4), 108-112 (2011-01-01)
The in vitro effects of two commercial sensitivity relief dentifrices, one containing 8.0% arginine, calcium carbonate, and 1450 ppm fluoride as sodium monofluorophosphate (MFP), and the other containing 8% strontium acetate and 1040 ppm fluoride as sodium fluoride, in occluding
Charles R Parkinson et al.
The Journal of clinical dentistry, 22(3), 74-81 (2011-09-13)
The objective of this study was to evaluate the relative level of dentin tubule occlusion and dentin mineralization conferred by a 5% w/w calcium sodium phosphosilicate (45S5)/1450 ppm fluoride toothpaste in comparison to a range of commercial toothpastes reported to

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service