Skip to Content
Merck
All Photos(3)

Documents

402850

Sigma-Aldrich

Calcium acetate monohydrate

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(CH3CO2)2 · H2O
CAS Number:
Molecular Weight:
176.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥99.0%

form

powder

impurities

≤0.005% insolubles
≤0.035 meq/g Titr. acid

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.05%
Na: ≤0.02%
Sr: ≤0.05%
heavy metals (as Pb): ≤0.005%

suitability

passes test for acidity or alkalinity

SMILES string

[H]O[H].CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium acetate monohydrate is calcium salt of acetic acid. Crystals study reveals the structure to be triclinic. Its solubility in aqueous solution over a range of temperature has been studied.

Application

Calcium acetate monohydrate may be used in the following processes:
  • As a calcium source in the preparation of calcium hydroxyapatite by sol–gel synthesis.
  • As a precursor in the synthesis of calcium oxide sorbent used in capturing CO2.
  • As a precursor salt in the synthesis of CaO/γ-Al2O3 ( calcium oxide/ aluminium oxide) heterogeneous base catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium oxide based sorbents for capture of carbon dioxide at high temperatures.
Hong L, et al.
Industrial & Engineering Chemistry Research, 45(11), 3944-3949 (2006)
The synthesis of CaO loaded onto Al2O3 from calcium acetate and its application in transesterification of the sunflower oil.
Marinkovic DM, et al.
Advanced technologies, 4(1) , 26-32 (2015)
Calcium hydroxyapatite, Ca10(PO4)6(OH)2 ceramics prepared by aqueous sol-gel processing.
Bogdanoviciene I, et al.
Materials Research Bulletin, 41(9), 1754-1762 (2006)
Structure of calcium acetate monohydrate, Ca(C2H3O2) 2.H2O.
Klop EA, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 40(1), 51-53 (1984)
Ash M and Ash I.
Handbook of Preservatives, 309-309 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service