Skip to Content
Merck
All Photos(1)

Documents

434191

Sigma-Aldrich

Vinyl bromide solution

1.0 M in THF

Synonym(s):

Bromoethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHBr
CAS Number:
Molecular Weight:
106.95
Beilstein:
1361370
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

12.46 psi ( 55 °C)
3.6 psi ( 20 °C)

Quality Level

form

liquid

concentration

1.0 M in THF

density

0.927 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

BrC=C

InChI

1S/C2H3Br/c1-2-3/h2H,1H2

InChI key

INLLPKCGLOXCIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vinyl bromide solution belongs to haloalkenes andis highly reactive due to the presence of an unsaturated vinyl group. It can provide flame retardancy to polymers or materials when incorporated into their structure. It is also a versatile building block for polymerization, addition reactions, substitution reactions, and cross-coupling reactions like Suzuki-Miyaura and Negishi reactions. It can be used to introduce radiolabel into molecules for medical imaging.

Application


  • Sequential Vinyl Radical Cyclization/Fixation of Carbon Dioxide through Electrochemical Reduction of Vinyl Bromide in the Presence of an Electron-Transfer Mediator: This study explores the electrochemical reduction of vinyl bromide with a focus on vinyl radical cyclization and carbon dioxide fixation (A Katayama, H Senboku, 2016).

  • A Comparison of the Wavelength-Dependent Photochemical Reactions of Ozone with Vinyl Bromide and Fluoride in Argon Matrices: The study compares the photochemical reactions of vinyl bromide and fluoride with ozone, examining their behavior in argon matrices (BS Ault, 2021).

Vinyl bromide solution can be used as a precursor for stereoselective synthesis of chiral 2-vinyl-tetrahydronaphthalens via asymmetric reductive coupling. These chiral compounds are valuable building blocks for natural products, agrochemicals, and liquid crystals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel/Copper Co-catalyzed Enantioselective Reductive Coupling of Oxabenzonorbornadienes with Vinyl Bromides
Yao Deng, et al.
advanced synthesis and catalysis, 365, 3265-3270 (2023)
Hsin-Lun Kao et al.
Organic letters, 13(19), 5204-5207 (2011-09-02)
The synthesis of vinyl sulfides through the coupling reaction of thiols with vinyl iodides, bromides, and chlorides is described. The thiols can couple with aryl iodides in the presence of only 0.5 mol % Cu(2)O without the need for an
Qiwu Zhao et al.
Organic letters, 10(18), 4037-4040 (2008-08-30)
A general and highly efficient synthesis of 4-alkylidene-2-azetidinones was achieved by the Cu(I)-catalyzed intramolecular C-N coupling of amides with vinyl bromides. This 4-exo ring closure was found to be fundamentally preferred over other modes (5-exo, 6-exo, and 6-endo) of cyclization
Changhui Sun et al.
Organic letters, 11(18), 4084-4087 (2009-08-20)
With the catalysis of CuI/trans-N,N'-dimethylcyclohexane-1,2-diamine, a number of carboxylic acids underwent efficient intramolecular O-vinylation with vinyl bromides leading to the synthesis of the corresponding five- and six-membered enol lactones. The same catalytic system also led to the efficient cycloisomerization of
Cheon-Gyu Cho et al.
Organic letters, 7(16), 3569-3572 (2005-07-29)
An effective, readily scalable two-step synthesis of trisubstituted (E)-vinyl bromides involving bromination of alpha,beta-unsaturated lactones followed by hydrolytic fragmentation has been developed. Several trisubstituted (E)-vinyl bromides, including multigram quantities of (+)-(E)-4-bromo-2-methyl-3-pentenol, a synthetic intermediate required for the C(8)-C(11) moieties of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service