Skip to Content
Merck
All Photos(1)

Key Documents

368679

Sigma-Aldrich

cis-1-Bromo-1-propene

97%

Synonym(s):

(Z)-1-Bromo-1-propene, (Z)-1-Bromoprop-1-ene, (Z)-1-Bromopropene, (Z)-1-Propenyl bromide, (Z)-Propenyl bromide, cis-1-Bromopropene, cis-1-Propenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHBr
CAS Number:
Molecular Weight:
120.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.4545 (lit.)

bp

58 °C (lit.)

density

1.423 g/mL at 25 °C (lit.)

SMILES string

C\C=C/Br

InChI

1S/C3H5Br/c1-2-3-4/h2-3H,1H3/b3-2-

InChI key

NNQDMQVWOWCVEM-IHWYPQMZSA-N

General description

cis-1-Bromo-1-propene is an aliphatic olefin. Oxidation of cis-1-bromo-1-propene by toluene dioxygenase has been reported.

Application

cis-1-Bromo-1-propene is used in the preparation of (Z)-1-lithio-1-propeneusing lithium powder.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of a functionalized 2-oxo-1-azabicyclo [5.3. 0] alkane as a potential scaffold for targeted chemotherapy strategies
Bracci A, et al.
Synthesis, 15, 2363-2367 (2003)
C C Lange et al.
Journal of bacteriology, 179(12), 3858-3865 (1997-06-01)
Toluene dioxygenase from Pseudomonas putida F1 has been studied extensively with aromatic substrates. The present work examined the toluene dioxygenase-catalyzed oxidation of various halogenated ethenes, propenes, butenes and nonhalogenated cis-2-pentene, an isomeric mix of 2-hexenes, cis-2-heptene, and cis-2-octene as substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service