434183
Magnesium sulfate monohydrate
97%
Synonym(s):
Kieserite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
MgSO4 · H2O
CAS Number:
Molecular Weight:
138.38
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030409
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Recommended Products
grade
for analytical purposes
Quality Level
Assay
97%
form
powder
mp
150 °C (dec.) (lit.)
density
2.57 g/mL at 25 °C (lit.)
SMILES string
O.[Mg++].[O-]S([O-])(=O)=O
InChI
1S/Mg.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2
InChI key
LFCFXZHKDRJMNS-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Magnesium sulfate monohydrate is a Lewis acid catalyst widely used in the field of organic synthesis, nanomaterial preparation, and fuel cells.
Application
Magnesium sulfate monohydrate can be used:
- As a precursor to synthesize magnetic manganese ferrite nanoparticles by alkaline coprecipitation.
- To fabricate manganese octahedral molecular sieve (OMS-2) catalysts for selective aerobic oxidation of aromatic and aliphatic thiols.
- As a catalyst to synthesize benzo-2-pyrones and benzopyrazines.
- As a precursor to synthesize manganese oxide nanoparticles for energy storage applications.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Manganese octahedral molecular sieve (OMS-2) catalysts for selective aerobic oxidation of thiols to disulfides
Saminda Dharmarathna, et al.},
Applied Catalysis. B, Environmental, 147, 124-131 (2014)
MnSO4. H2O: A highly efficient and inexpensive catalyst for the synthesis of benzo-2-pyrones and benzopyrazines
F Jafari and S Khodabakhshi
Bulgarian Chemical Communications, 46, 36-42 (2014)
Influence of Manganese Dioxide Nanoparticles on MoS2/PANI Nanosheets and Its Energy Storage Applications
S Krithika and J Balavijayalakshmi
Journal of Inorganic and Organometallic Polymers and Materials, 1-10 (2023)
Biotinylated magnetic nanoparticles for pretargeting: synthesis and characterization study
Cancer Nanotechnology, 2, 111-120 (2011)
Ricardo Arevalo et al.
Rapid communications in mass spectrometry : RCM, 32(21), 1875-1886 (2018-07-27)
The investigation of cryogenic planetary environments as potential harbors for extant life and/or contemporary sites of organic synthesis represents an emerging focal point in planetary exploration. Next generation instruments need to be capable of unambiguously determining elemental and/or molecular stoichiometry
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service