Skip to Content
Merck
All Photos(1)

Key Documents

450073

Sigma-Aldrich

Ytterbium(III) chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Ytterbium chloride, Ytterbium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbCl3
CAS Number:
Molecular Weight:
279.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤150.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

875 °C (lit.)

SMILES string

Cl[Yb](Cl)Cl

InChI

1S/3ClH.Yb/h3*1H;/q;;;+3/p-3

InChI key

CKLHRQNQYIJFFX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Albin et al.
Journal of animal science, 79(5), 1250-1258 (2001-05-26)
The apparent ileal digestibilities of amino acids and rate of passage were evaluated in pigs (BW = 78.3 +/- 7.4 kg) fed a semipurified diet. The pigs were fed 1.82, 2.73, or 3.65 kg DMI/d. The highest level of feed
Zhang, Y. et al.
Synthetic Communications, 27, 4327-4327 (1997)
W Shepard et al.
Acta crystallographica. Section D, Biological crystallography, 56(Pt 10), 1288-1303 (2000-09-22)
The basic theory and principles of the multiple-wavelength anomalous solvent-contrast (MASC) method are introduced as a contrast-variation technique for generating low-resolution crystallographic phase information on the envelope of a macromolecule. Experimental techniques and practical considerations concerning the choice of anomalous
Yuji Ohashi et al.
International journal of food microbiology, 96(1), 61-66 (2004-09-11)
The transition of probiotic bacteria Lactobacillus in the gastrointestinal tract was investigated in pigs that received commercially available fermented milk prepared with Lactobacillus casei strain Shirota (LCS). Three female pigs fistulated at the cecum were fed 130 ml of fermented
Haiqing Xiao et al.
Toxicology letters, 155(2), 247-252 (2004-12-18)
It is well known that lanthanides have come into extensive and rapid use in a number of fields. As a result, more and more lanthanides are getting into the environment and food chains. However, the distributions of lanthanides in brain

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service