Skip to Content
Merck
All Photos(3)

Key Documents

449857

Sigma-Aldrich

Lanthanum(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Lanthanum fluoride, Lanthanum trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaF3
CAS Number:
Molecular Weight:
195.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤200.0 ppm Trace Rare Earth Analysis

density

5.936 g/mL at 25 °C (lit.)

SMILES string

F[La](F)F

InChI

1S/3FH.La/h3*1H;/q;;;+3/p-3

InChI key

BYMUNNMMXKDFEZ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Employed extensively in the preparation and study of fluoride glasses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Laure Rollet et al.
Physical chemistry chemical physics : PCCP, 10(22), 3222-3228 (2008-05-27)
The local structures of molten lanthanum alkali fluoride binaries have been studied using HT NMR technique. The chemical shifts of (19)F, (23)Na and (139)La in solid and in liquid have been compared for AF (A = alkali) and LaF(3). In
Journal of Materials Science Letters, 15, 21-21 (1996)
Sri Sivakumar et al.
Journal of the American Chemical Society, 127(36), 12464-12465 (2005-09-08)
White light was generated from a single silica thin film made with Yb0.75La0.2Eu0.05F3, La0.45Yb0.5Er0.05F3, and La0.75Yb0.2Tm0.05F3 nanoparticles by exciting with a single source near-infrared light (980 nm CW diode laser). Eu3+ and Tm3+ ions are responsible for red and blue
Radiative lifetime modification of LaF₃:Nd nanoparticles embedded in 3D silicon photonic crystals.
Hailong Ning et al.
Advanced materials (Deerfield Beach, Fla.), 24(23), OP153-OP158 (2012-05-10)
Daibo Pi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(11-12), 2455-2459 (2005-07-27)
Europium-doped LaF3 nanoparticles have been prepared by the ionic reaction in the ethanol at 60 degrees C. From the XRD pattern of nanoparticles and the emission spectra of Eu3+ ions, it has been concluded that the Eu3+ ions could easily

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service