422037
DBE-3
Synonym(s):
Dibasic ester mixture, Mixture of dimethyl adipate and dimethyl glutarate
About This Item
Recommended Products
vapor pressure
0.2 mmHg ( 20 °C)
Quality Level
description
mixture of dimethyl adipate and dimethyl glutarate
form
liquid
autoignition temp.
698 °F
expl. lim.
8 %
refractive index
n20/D 1.428 (lit.)
bp
215-225 °C (lit.)
density
1.068 g/mL at 25 °C (lit.)
SMILES string
COC(=O)CCCC(=O)OC.COC(=O)CCCCC(=O)OC
InChI
1S/C8H14O4.C7H12O4/c1-11-7(9)5-3-4-6-8(10)12-2;1-10-6(8)4-3-5-7(9)11-2/h3-6H2,1-2H3;3-5H2,1-2H3
InChI key
FSCIDASGDAWVED-UHFFFAOYSA-N
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
215.6 °F
Flash Point(C)
102 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service