Skip to Content
Merck
All Photos(3)

Documents

112755

Sigma-Aldrich

DBE-4 dibasic ester

98%

Synonym(s):

Dimethyl succinate, Succinic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH2COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein:
956776
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.3 mmHg ( 20 °C)

Assay

98%

autoignition temp.

689 °F

expl. lim.

8.5 %

refractive index

n20/D 1.419 (lit.)

bp

200 °C (lit.)

mp

16-19 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl succinate (DBE-4 dibasic ester ) is obtained by the oxidation of 1,4-butanediol and butyrolactone by using supported gold, palladium and gold-palladium nanoparticles.

DBE-4 can be used as an intermediate in the preparation of 1,4-butanediol, γ-butyrolactone, tetrahydrofuran and itaconic acid.

Legal Information

DuPont product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma L Brett et al.
ChemSusChem, 6(10), 1952-1958 (2013-10-10)
The oxidation of 1,4-butanediol and butyrolactone have been investigated by using supported gold, palladium and gold-palladium nanoparticles. The products of such reactions are valuable chemical intermediates and, for example, can present a viable pathway for the sustainable production of polymers.
Determination and correlation of ternary isobaric vapour-liquid equilibrium data of (dimethyl succinate+ dimethyl glutarate+ dimethyl adipate) at 2, 5 and 8 kPa
Zhi C, et al.
The Journal of Chemical Thermodynamics, 143, 106047-106047 (2020)
V Leclercq-Meyer et al.
Biochemical and molecular medicine, 59(1), 87-90 (1996-10-01)
Glucagon-like peptide 1 (GLP-1) is often referred to as a glucose-dependent insulinotropic agent and is currently under investigation as a tool in the treatment of noninsulin-dependent diabetes. This report shows that, in the absence of glucose, a nonglucidic nutrient, namely
V Leclercq-Meyer et al.
Life sciences, 58(14), 1195-1199 (1996-01-01)
The glucagon-like peptide 1 (7-36) amide (GLP-1, 1.0 nM) was administered to isolated rat pancreases perfused either in the absence of exogenous nutrient or presence of 10 mM succinic acid dimethyl ester (SAD). In the absence of any exogenous nutrient
L Ladrière et al.
Metabolism: clinical and experimental, 48(1), 102-106 (1999-01-27)
The metabolism of [2,3-13C]succinic acid dimethyl ester ([2,3-13C]-SAD) 10 mmol/L was examined in hepatocytes from overnight-fasted normal rats, 3-day starved rats, and overnight-fasted hereditarily diabetic Goto-Kakizaki (GK) rats. The amount of 13C-labeled succinate, fumarate, malate, lactate, alanine, and aspartate released

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service