Skip to Content
Merck
All Photos(3)

Key Documents

412244

Sigma-Aldrich

2-Fluorobenzoic acid

97%

Synonym(s):

o-Fluorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO2H
CAS Number:
Molecular Weight:
140.11
Beilstein:
971265
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

122-125 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccccc1F

InChI

1S/C7H5FO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

InChI key

NSTREUWFTAOOKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victor L Rendina et al.
The Journal of organic chemistry, 77(2), 1181-1185 (2012-01-14)
A new protocol for titrating nonstabilized diazoalkane solutions by quantitative (19)F NMR is reported. An excess of 2-fluorobenzoic acid dissolved in CDCl(3) is treated with the diazoalkane solution at a low temperature, immediately forming the corresponding 2-fluorobenzoate ester upon warming.
O Drzyzga et al.
FEMS microbiology letters, 116(2), 215-219 (1994-02-15)
A mesophilic, dehalogenating, sulfate-reducing diculture was isolated from an anaerobic lake sediment. One strain of the diculture is proposed to be an endospore-forming Desulfotomaculum species, the second strain was a vibrioid, motile and non-sporeforming species which is tentatively assigned to
S Y Lee et al.
Nuclear medicine and biology, 28(4), 391-395 (2001-06-08)
In vitro metabolism of acetylcholinesterase inhibitors containing 3-[(18)F]fluoromethylbenzyl- ([(18)F]1) and 4-[(18)F]fluorobenzyl-piperidine moieties ([(18)F]2) was studied and compared with the in vivo metabolism. Defluorination of the [(18)F]1 mainly occurred to generate [(18)F]fluoride ion both in vitro and in vivo. In contrast
M Begoña Osuna et al.
Water research, 42(14), 3857-3869 (2008-07-29)
Two up-flow fixed-bed reactors (UFBRs), inoculated with activated sludge and operated for 162 days, were fed 1mmolL(-1)d(-1) with two model halogenated compounds, 2-fluorobenzoate (2-FB) and dichloromethane (DCM). Expanded clay (EC) and granular activated carbon (GAC) were used as biofilm carrier.
U Schennen et al.
Journal of bacteriology, 161(1), 321-325 (1985-01-01)
Three strains of anaerobically benzoate-degrading, denitrifying bacteria of the genus Pseudomonas were able to grow on 2-fluorobenzoate as the sole carbon and energy source. Fluoride ion release was stoichiometric, and the reduction of dissolved organic carbon indicated total degradation. Cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service