128384
4-Fluorobenzoic acid
98%
Synonym(s):
p-Fluorobenzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
FC6H4CO2H
CAS Number:
Molecular Weight:
140.11
Beilstein:
1906922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
182-184 °C (lit.)
solubility
alcohol: soluble
cold water: very slightly soluble
hot water: freely soluble
functional group
carboxylic acid
fluoro
SMILES string
OC(=O)c1ccc(F)cc1
InChI
1S/C7H5FO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
InChI key
BBYDXOIZLAWGSL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Fluorobenzoic acid has been used in high resolution charge density study.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jan Marik et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(2), 199-203 (2006-08-29)
The widely used bifunctional labeling reagent 4-[18F]fluorobenzoic acid ([18F]FBA) and its activated form N-succinimidyl 4-[18F]fluorobenzoate ([18F]SFB) were prepared using a modified Siemens/CTI chemistry process control unit (CPCU) double vessel [18F]FDG module. The products were obtained with a radiochemical yield greater
Julien Andriès et al.
Bioorganic & medicinal chemistry letters, 20(12), 3730-3733 (2010-05-18)
In search of a serotonin 5-HT(7) radiotracer for positron emission tomography, we developed 1-[2-[(2S)-1-(phenylsulfonyl)pyrrolidin-2-yl]ethyl]piperidin-4-yl 4-fluorobenzoate. After labeling in good yield with fluorine-18 via nitro for fluorine exchange, preliminary biological experiments with autoradiographies failed to evidence any specific 5-HT(7) receptor delineation.
J L Sutcliffe-Goulden et al.
Bioorganic & medicinal chemistry letters, 10(14), 1501-1503 (2000-07-29)
A strategy for the solid phase synthesis of [18F]labelled peptides has been developed. The peptides were prepared on solid support and acylated with 4-[18F]fluorobenzoic acid using HATU within 3 min and the labelled peptide was released from the solid support
Comparison of [18F]FBA and [18F]FPyMe as peptide radiolabeling agents of PEPHC1 for PET imaging of EGFRvIII.
Charlotte L Hansen et al.
Advances in experimental medicine and biology, 611, 405-406 (2009-04-30)
Benjamin J Hackel et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(7), 1101-1105 (2013-05-15)
Integrin αvβ6 is a cell surface receptor minimally expressed by healthy tissue but elevated in lung, colon, skin, ovarian, cervical, and pancreatic cancers. A molecular PET agent for integrin αvβ6 could provide significant clinical utility by facilitating both cancer staging
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service