Skip to Content
Merck
All Photos(1)

Documents

P5360

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzoic acid

99%

Synonym(s):

pentafluorobenzoic acid, perfluorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CO2H
CAS Number:
Molecular Weight:
212.07
Beilstein:
2054395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

220 °C (lit.)

mp

100-102 °C (lit.)

SMILES string

OC(=O)c1c(F)c(F)c(F)c(F)c1F

InChI

1S/C7HF5O2/c8-2-1(7(13)14)3(9)5(11)6(12)4(2)10/h(H,13,14)

InChI key

YZERDTREOUSUHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasan Muhammad Zubair et al.
Journal of plant research, 130(4), 647-658 (2017-03-24)
Alfalfa (Medicago sativa L.) genotypes at varying densities were investigated for allelopathic impact using annual ryegrass (Lolium rigidum) as the target species in a laboratory bioassay. Three densities (15, 30, and 50 seedlings/beaker) and 40 alfalfa genotypes were evaluated by
Gina B Barbosa et al.
Phytochemistry, 140, 141-150 (2017-05-13)
Zingiberaceae plants, commonly known as gingers, have been popular for their medicinal and culinary uses since time immemorial. In spite of their numerous health-promoting applications, many Zingiberaceae plants still receive no scientific attention. Moreover, existing reports mostly focused only on
Jiann-Ming Wang et al.
Environmental toxicology and chemistry, 24(11), 2806-2811 (2006-01-10)
The biodegradation and transport of phenanthrene in porous media containing multiple populations of phenanthrene degraders is examined with a series of miscible-displacement experiments. A long-term experiment was conducted with a soil containing an indigenous microbial community comprised of 25 identified
Rohit Salve
Ground water, 43(1), 133-137 (2005-02-25)
This paper presents the design of the passive-discrete water sampler (PDWS) which has been developed to facilitate investigations of flow partitioning in fractured rocks. The PDWS continuously isolates seeping water into discrete samples while monitoring the seepage rate. The PDWS
Sonnich Meier et al.
Journal of chromatography. A, 1062(2), 255-268 (2005-02-01)
A highly selective and sensitive method for the determination of 30 meta- and para-substituted alkylphenols from phenol (C0) to nonylphenol (C9) in biota is described. Dichloromethane extracts of spiked cod liver and muscle samples are cleaned up by gel permeation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service