Skip to Content
Merck
All Photos(1)

Key Documents

88908

Sigma-Aldrich

Methoxypolyethylene glycol 5,000 propionic acid

≥80%

Synonym(s):

Polyethylene glycol, O-(2-Carboxyethyl)-O′-methylpolyethylene glycol 5,000, mono-Methyl polyethylene glycol 5′000 propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25

Assay

≥80%

form

lumps

Ω-end

carboxylic acid

α-end

methoxy

storage temp.

−20°C

Application

Methoxypolyethylene glycol 5,000 propionic acid (M-SPA-PEG 5000) is a pegylation agent used to conjugate proteins to reduce immunogenicity and prolong serum half-life of proteins and enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihong Huang et al.
International journal of hematology, 92(4), 609-616 (2010-10-19)
We designed to determine whether pretreatment of allografts with methoxypolyethylene glycol-succinimidyl-propionic acid ester (mPEG-SPA) and an anti-OX40L monoclonal antibody (McAb) can relieve acute graft-versus-host disease in allogeneic bone marrow transplantation recipients. Responder splenocytes from C57BL/6 donor mice were incubated with
Y Tan et al.
Protein expression and purification, 12(1), 45-52 (1998-03-14)
Recombinant methioninase (rMETase) is a homotetrameric pyridoxal 5'-phosphate enzyme of 172-kda molecular mass derived from Pseudomonas putida and cloned in Escherichia coli. rMETase has been found previously to be an effective, anti-tumor agent in vitro and in vivo. The enzyme
Amanda J Bradley et al.
Biochimica et biophysica acta, 1561(2), 147-158 (2002-05-09)
Immunocamouflaged red blood cells (RBC) are produced by cell surface derivatization with methoxypolyethylene glycol (mPEG). These immunologically attenuated cells may reduce the risk of allosensitization in chronically transfused patients. To characterize the effects of differing linker chemistries and polymer lengths
Chun Zhang et al.
PloS one, 7(6), e39659-e39659 (2012-06-30)
Uricase has proven therapeutic value in treating hyperuricemia but sufficient reduction of its immunogenicity may be the largest obstacle to its chronic use. In this study, canine uricase was modified with 5 kDa mPEG-SPA and the impact of large aggregated

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service