Skip to Content
Merck
All Photos(1)

Documents

407275

Sigma-Aldrich

N,N-Dimethylaniline

purified by redistillation, ≥99.5%

Synonym(s):

N,N-Dimethylaniline, N,N-Dimethylphenylamine, DMA, Dimethylaniline, Dimethylphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.5%

form

liquid

purified by

glass distillation
redistillation

refractive index

n20/D 1.557 (lit.)

pH

7.4 (20 °C, 1.2 g/L)

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anodic oxidation of N,N-dimethylaniline at platinum electrode in acidic buffers has been studied by conventional and cyclic voltammetry. Photoreduction of excited benzophenone (BP) by DMA in acetonitrile solution has been investigated by picosecond-femtosecond laser photolysis and time resolved transient absorption spectroscopy.

Application

N,N-Dimethylaniline (DMA) may be used for the preparation of poly(N,N-dimethylaniline) film by anodic oxidation of DMA.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemically polymerized N, N-dimethylaniline film with ion-exchange properties as an electrode modifier.
Oyama N, et al.
Analytical Chemistry, 57(8), 1526-1532 (1985)
Femtosecond-picosecond laser photolysis studies on photoreduction process of excited benzophenone with N,N-dimethylaniline in acetonitrile solution.
Miyasaka H, et al.
Bulletin of the Chemical Society of Japan, 63(12), 3385-3397 (1990)
Anodic oxidation studies of N,N-dimethylaniline. I. Voltammetric and spectroscopic investigations at platinum electrodes.
Mizoguchi T and Adams RN.
Journal of the American Chemical Society, 84(11), 2058-2061 (1962)
Yong Wang et al.
The journal of physical chemistry. B, 114(8), 2964-2970 (2010-02-12)
This paper addresses the experimentally observed mechanistic differences between the cytochrome P450-catalyzed N-demethylation of substituted N,N-dimethylanilines (DMA) and of N,N-dimethylbenzamides (DMBA). The two reactions of these substrates are initiated by C-H activation of the methyl groups on the nitrogen. Thus
Marius Koch et al.
Journal of the American Chemical Society, 134(8), 3729-3736 (2012-01-31)
The fluorescence quenching of 3-cyanoperylene upon electron transfer from N,N-dimethylaniline in three room-temperature ionic liquids (RTILs) and in binary solvent mixtures of identical viscosity has been investigated using steady-state and time-resolved fluorescence spectroscopy. This study was stimulated by previous reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service