Skip to Content
Merck
All Photos(1)

Key Documents

D145750

Sigma-Aldrich

N,N-Dimethylaniline

99%

Synonym(s):

N,N-Dimethylphenylamine, DMA, Dimethylaniline, Dimethylphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.557 (lit.)

bp

193-194 °C (lit.)

mp

1.5-2.5 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)c1ccccc1

InChI

1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

JLTDJTHDQAWBAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-dimethylaniline (DMA) is N,N-disubstituted aniline. The thermo-physical and thermodynamic properties of the binary mixtures of DMA with anisole (ANS) and tert-butyl methyl ether (MTBE) have been reported.[1] In acidic buffers it forms N,N,N′,N′-tetramethylbenzidine at a platinum electrode due anodic oxidation.[2] DMA combines with diazonium salts to form methyl orange under alkaline condition.[3] It forms ortho- and para-nitro derivatives via nitration reaction using N-bromosuccinimide/silver(I) nitrate reagent system in acetonitrile.[4]

Application

N,N-dimethylaniline (DMA) may be used as a catalyst for reductive lithiation.[5]
Reagent in new methodology for symmetrical and unsymmetrical photoconductor squaranes.[6]

Legal Information

DuPont product

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fundamental Difference in Reductive Lithiations with Preformed Radical Anions versus Catalytic Aromatic Electron-Transfer Agents: N,N-Dimethylaniline as an Advantageous Catalyst.
Kennedy N, et al.
Angewandte Chemie (International Edition in English), 128(1), 391-394 (2016)
Spectrophotometric determination of chromium by using sulphanilic acid and N, N-dimethylaniline.
Chandrashekhara KG, et al.
Indian Journal of Chemical Technology, 22(1-2), 78-81 (2015)
Aromatic nitration under neutral conditions using N-bromosuccinimide/silver (I) nitrate.
Nowrouzi N, et al.
Tetrahedron Letters, 53(36), 4841-4842 (2012)
Canadian Journal of Chemistry, 71, 494-494 (1993)
Anodic oxidation studies of N,N-dimethylaniline. I. Voltammetric and spectroscopic investigations at platinum electrodes.
Mizoguchi T and Adams RN.
Journal of the American Chemical Society, 84(11), 2058-2061 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service