Skip to Content
Merck
All Photos(4)

Key Documents

471216

Sigma-Aldrich

Diethylamine

≥99.5%

Synonym(s):

N,N-Diethylamine, N-ethyl-ethanamine, DEA, Diethamine, Diethyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
technique(s):
GC/MS: suitable
bp:
55 °C (lit.)
vapor pressure:
14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

vapor density

2.5 (vs air)

Quality Level

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

594 °F

expl. lim.

10.1 %

technique(s)

GC/MS: suitable

refractive index

n20/D 1.385 (lit.)

pH

13 (20 °C, 100 g/L)

bp

55 °C (lit.)

mp

−50 °C (lit.)

solubility

water: soluble

density

0.707 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylamine (DEA) is an organic base and a weak σ-donor ligand.

Application

Diethylamine has been used to constitute the extraction solvent mixture for the gas chromatography-mass spectrometric (GC-MS) analysis of melamine and related compounds. It has been also been used as a bond breaking agent in a novel ELISA technique to measure the avidity index (AI) of IgG-anti-PT (Immunoglobulin G-anti-pertussis toxin) antibodies.
It may be used as:
  • An organocatalyst for the preparation of β-phosphonomalononitriles and diethyl (2-amino-3- cyano-chromene-4-yl) phosphonic acid esters.
  • A ligand for the synthesis of (N-heterocyclic carbene)PdCl2(diethylamine) complexes.
  • A template for the preparation of thermally and hydrothermally stable SAPO-34 (silicoaluminophosphate) molecular sieves.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and catalytic properties of SAPO-34 synthesized using diethylamine as a template.
Liu G, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 111(1), 143-149 (2008)
Alex-Mikael Barkoff et al.
Vaccine, 30(48), 6897-6902 (2012-09-18)
Pertussis toxin (PT) is a specific virulence factor of Bordetella pertussis and it is included in all acellular pertussis vaccines (aP). Although immunity after infection seems to persist longer than that after vaccination, the exact mechanism(s) is not known. Primary
A practical and highly efficient protocol for multicomponent synthesis of β-phosphonomalononitriles and 2-amino-4H-chromen-4-yl phosphonates using diethylamine as a novel organocatalyst.
Kulkarni MA, et al.
Comptes Rendus Chimie, 15(9), 745-752 (2012)
Inhibited Catalyst Activation in (N-Heterocyclic carbene) PdCl2 (diethylamine) Complexes by Intramolecular Hydrogen Bonding.
Chen MT, et al.
Organometallics, 30(24), 6770-6773 (2011)
Tongyao Wei et al.
STAR protocols, 2(3), 100777-100777 (2021-09-07)
Dissecting the function of proteins' post-translational modifications (PTMs) is seriously hindered by the difficulty in obtaining the homogeneous protein with the PTMs of interest. Chemical protein synthesis offers a great potential to overcome this limitation. Here, a detailed protocol is

Articles

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service